Skip to Content
Merck
All Photos(1)

Documents

AB5087

Sigma-Aldrich

Anti-Melanocyte Stimulating Hormone α Antibody

serum, Chemicon®

Synonym(s):

alpha-MSH, POMC, Pro-opiomelanocortin, Corticotropin-lipotropin precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

sheep

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity (predicted by homology)

mammals

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
radioimmunoassay: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... POMC(5443)

Specificity

Specific for alpha-melanocyte stimulating hormone (aMSH), C-terminal.

Immunogen

MSH conjugated with bovine thyroglobulin.

Application

Anti-Melanocyte Stimulating Hormone Antibody, α detects level of Melanocyte Stimulating Hormone & has been published & validated for use in IH, RIA.
Immunohistochemistry: 1:10,000-1:50,000 on 4% paraformaldehyde fixed tissue.

RIA: 1:250,000-1:300,000.

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
CNS Control of Metabolism

Hormones & Receptors

Physical form

Sheep Serum. Liquid. Contains no preservative.
Unpurified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Hypothalamus

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maja Cigrovski Berkovic et al.
Neuroendocrinology, 99(2), 75-84 (2014-04-02)
Although previously considered rare, recent epidemiological studies have revealed that the incidence (3.6/100,000) and prevalence (35/100,000) of gastroenteropancreatic neuroendocrine tumors (GEP-NETs) has increased over the past few decades. Despite the progress in the understanding of GEP-NET molecular biology, there is
Fumihiko Maekawa et al.
Frontiers in synaptic neuroscience, 5, 7-7 (2013-10-10)
We previously reported that the type 2 diabetic Goto-Kakizaki (GK) rats at young adult ages (6-12 weeks) exhibited increased visceral fat mass and hyperleptinemia, due to hyperphagia caused primarily by neuropeptide Y (NPY) overexpression in the hypothalamic arcuate nucleus. Later
B E Grayson et al.
Endocrinology, 151(4), 1622-1632 (2010-02-24)
The hypothalamic melanocortin system, which controls appetite and energy expenditure, develops during the third trimester in primates. Thus, maternal nutrition and health may have a profound influence on the development of this system. To study the effects of chronic maternal
Amanda H Mortensen et al.
Human molecular genetics, 24(4), 939-953 (2014-10-16)
OTX2 is a homeodomain transcription factor that is necessary for normal head development in mouse and man. Heterozygosity for loss-of-function alleles causes an incompletely penetrant, haploinsufficiency disorder. Affected individuals exhibit a spectrum of features that range from developmental defects in
Processing and sorting of the prohormone convertase 2 propeptide.
Muller, L; Cameron, A; Fortenberry, Y; Apletalina, EV; Lindberg, I
The Journal of Biological Chemistry null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service