Skip to Content
Merck
All Photos(1)

Documents

AB3521

Sigma-Aldrich

Anti-Leptin Antibody

Chemicon®, from chicken

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

chicken

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... LEP(3952)

Related Categories

Specificity

Leptin, rat and mouse.

Immunogen

Recombinant rat leptin.

Application

Detect Leptin using this Anti-Leptin Antibody validated for use in ELISA & WB.
Immunoblotting: 1:1,000-1:5,000

ELISA: 1:1,000-1:5,000

Optimal working dilutions must be determined by the end user.
Research Category
Metabolism
Research Sub Category
Lipid Metabolism & Weight Regulation

Linkage

Replaces: MAB3870

Physical form

Affinity purified immunoglobulin. Liquid in PBS containing 0.02% sodium azide.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bo-Mi Kim et al.
International journal of molecular medicine, 43(1), 489-500 (2018-10-27)
Muscat Bailey A grape stalk is an organic waste produced in marked amounts during the vinification of grapes. Previous studies have indicated that grape stalk is rich in bioactive phenolic compounds, and exhibits antioxidant and UV‑protective activities. However, its effects
Jiung-Pang Huang et al.
Journal of molecular endocrinology, 60(4), 307-321 (2018-03-28)
Many studies have reported the causes of obese metabolic syndrome (MS); however, the causes of nonobese MS (NMS) remain unknown. In this study, we demonstrated that inflamed dysfunctional adipose tissue plays a crucial role in cholesterol-induced NMS. Control (C), high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service