Skip to Content
Merck
All Photos(2)

Key Documents

AB2237

Sigma-Aldrich

Anti-PAX6 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Aniridia type II protein, paired box 6, paired box gene 6, paired box gene 6 (aniridia, keratitis), paired box homeotic gene-6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, rat, chicken

should not react with

human

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PAX6(5080)

General description

PAX6 is the most researched of the PAX genes and appears throughout the literature as a "master control" gene for the development of eyes and other sensory organs, certain neural and epidermal tissues as well as other homologous structures, usually derived from ectodermal tissues. This transcription factor is most famous for its use in the interspecifically induced expression of ectopic eyes and is of medical importance because heterozygous mutants produce a wide spectrum of ocular defects such as Aniridia in humans. This gene encodes paired box gene 6, one of many human homologues of the Drosophila melanogaster gene prd. In addition to the hallmark feature of this gene family, a conserved paired box domain, the encoded protein also contains a homeo box domain. Both domains are known to bind DNA, and function as regulators of gene transcription. This gene is expressed in the developing nervous system, and in developing eyes. Mutations in this gene are known to cause aniridia as well as Peter′s anomaly, both ocular diseases.

Specificity

This antibody recognizes the C-Terminus of PAX6.

Immunogen

KLH conjugated followed by PAX6 corresponding to the C-Terminus

Application

Anti-PAX6 Antibody detects level of PAX6 & has been published & validated for use in WB & IC.
Immunocytochemistry:
Working dilutions of 1:50 - 1:1,000 of Anti-Pax6 was used to resolve the protein in chicken spinal cord.

Quality

Routinely evaluated by Western Blot on rat and mouse brain lysates.

Western Blot Analysis:
0.5 µg/mL of this lot detected PAX6 on 10 µg of mouse and rat brain lysates.

Target description

~48 kDa

Linkage

Replaces: AB5409

Physical form

Immunoaffinity purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH7.4) 150mM NaCl with 0.05% NaN3.

Analysis Note

Control
Mouse and rat brain lysates

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keith W Vance et al.
The EMBO journal, 33(4), 296-311 (2014-02-04)
Although some long noncoding RNAs (lncRNAs) have been shown to regulate gene expression in cis, it remains unclear whether lncRNAs can directly regulate transcription in trans by interacting with chromatin genome-wide independently of their sites of synthesis. Here, we describe
Erin Knock et al.
Neural development, 10, 13-13 (2015-05-03)
Chromatin-modifying complexes have key roles in regulating various aspects of neural stem cell biology, including self-renewal and neurogenesis. The methyl binding domain 3/nucleosome remodelling and deacetylation (MBD3/NuRD) co-repressor complex facilitates lineage commitment of pluripotent cells in early mouse embryos and
Sanghee Yun et al.
Development (Cambridge, England), 136(23), 3895-3906 (2009-11-13)
A crucial step in eye organogenesis is the transition of the optic vesicle into the optic cup. Several transcription factors and extracellular signals mediate this transition, but whether a single factor links them into a common genetic network is unclear.
Abby L Manthey et al.
Investigative ophthalmology & visual science, 55(8), 5445-5455 (2014-08-02)
Posterior capsular opacification (PCO), the most prevalent side effect of cataract surgery, occurs when residual lens epithelial cells (LECs) undergo fiber cell differentiation or epithelial-to-mesenchymal transition (EMT). Here, we used a murine cataract surgery model to investigate the role of
Noriaki Sasai et al.
PLoS biology, 12(7), e1001907-e1001907 (2014-07-16)
A relatively small number of signals are responsible for the variety and pattern of cell types generated in developing embryos. In part this is achieved by exploiting differences in the concentration or duration of signaling to increase cellular diversity. In

Articles

Human iPSC neural differentiation media and protocols used to generate neural stem cells, neurons and glial cell types.

Protocols

Step-by-step culture protocols for neural stem cell culture including NSC isolation, expansion, differentiation and characterization.

Step-by-step culture protocols for neural stem cell culture including NSC isolation, expansion, differentiation and characterization.

Step-by-step culture protocols for neural stem cell culture including NSC isolation, expansion, differentiation and characterization.

Step-by-step culture protocols for neural stem cell culture including NSC isolation, expansion, differentiation and characterization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service