Skip to Content
Merck
All Photos(1)

Documents

AB19012

Sigma-Aldrich

Anti-Laminin Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Specificity

This product has been prepared by immunoaffinity chromatography using immobilized antigens followed by extensive cross-adsorption against human serum proteins collagen and non-collagen extracellular matrix proteins to remove unwanted reactivities. Typically less than 1% cross reactivity against other extracellular matrix proteins was detected by ELISA against purified standards. This antibody reacts with most mammalian laminins and has negligible cross-reactivity with Type I, II, III, IV, V or VI collagens or fibronectin. Non-specific cross reaction of anti-laminin antibodies with other human serum proteins or non-laminin extracellular matrix proteins is negligible.

Immunogen

Full length human laminin. Immunization occurred after single-band purity was assessed by SDS-PAGE.

Application

Detect Laminin using this Anti-Laminin Antibody validated for use in ELISA, IP, WB, IH.
Immunoblot: 1:500-1:2000

ELISA: 1:4000-1:8000

Immunohistochemistry: 1:50-1:200

Immunoprecipitation: Condition must be determined by the end user

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified immunoglobulin. Liquid at 1 mg/mL in 0.125 M Sodium Borate, 0.075M Sodium Chloride, 0.005M EDTA, pH 8.0 with 0.01% Sodium Azide.
Format: Purified

Storage and Stability

The undiluted antibody solution is stable for approximately 6 months stored between 2 and 8° C. For extended storage mix with an equal volume of glycerol, aliquot and store at -20°C. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dhruv Sareen et al.
PloS one, 4(10), e7630-e7630 (2009-11-10)
Stem cell expansion and differentiation is the foundation of emerging cell therapy technologies. The potential applications of human neural progenitor cells (hNPCs) are wide ranging, but a normal cytogenetic profile is important to avoid the risk of tumor formation in
Extracellular matrix-mediated membrane-type 1 matrix metalloproteinase expression in pancreatic ductal cells is regulated by transforming growth factor-beta1.
Ottaviano, AJ; Sun, L; Ananthanarayanan, V; Munshi, HG
Cancer Research null
Cathepsin-L influences the expression of extracellular matrix in lymphoid organs and plays a role in the regulation of thymic output and of peripheral T cell number.
Gabriela Lombardi, Dalia Burzyn, Juliana Mundi?ano, Paula Berguer, Pedro Bekinschtein et al.
Journal of immunology (Baltimore, Md. : 1950) (1950)
Notch3 null mutation in mice causes muscle hyperplasia by repetitive muscle regeneration.
Takeo Kitamoto,Kazunori Hanaoka
Stem Cells null
Selective matrix metalloproteinase inhibition with developing heart failure: effects on left ventricular function and structure.
Mary K King, Mytsi L Coker, Aaron Goldberg, James H McElmurray, Himali R Gunasinghe et al.
Circulation Research null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service