Tenascin is known to play an active role in development of central nervous system and mesenchymal-derived organs, exist in the vasculature of tumors in adults, and function in cell adhesion.
Specificity
Reactivity with other species has not been confirmed, but likely to react with all mammalian species.
Reacts strongly with tenascin from human and mouse in both Western blotting and immunohistochemistry. While it has not been tested, it should also react strongly in these applications with tenascin from all mammalian species.
Immunogen
Purified human tenascin from glioma culture supernatant.
Application
Research Category Cell Structure
Research Sub Category ECM Proteins
This Anti-Tenascin Antibody is validated for use in WB, IH(P) for the detection of Tenascin.
Western blot: 0.5 - 1.0 μg/mL; recognizes 250-350 kDa polypeptides in reduced samples. Immunohistochemistry: 10 to 20 μg/mL in paraffin embedded tissues
Optimal working dilutions must be determined by end user.
Physical form
Affinity purified using immobilized human tenascin; presented as a liquid in PBS containing no preservatives.
Storage and Stability
Maintain at -20°C for up to 12 months from date of receipt. Aliquot to avoid repeated freeze/thaw cycles.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
: Although cancer stem cells (CSC) are thought to be responsible for tumor recurrence and resistance to chemotherapy, CSC-related research and drug development have been hampered by the limited supply of diverse, patient-derived CSC. Here, we present a functional polymer
International journal of molecular sciences, 22(6) (2021-04-04)
SARS-CoV-2 infection can cause cytokine storm and may overshoot immunity in humans; however, it remains to be determined whether virus-induced soluble mediators from infected cells are carried by exosomes as vehicles to distant organs and cause tissue damage in COVID-19
Loss of caveolin-1 from bronchial epithelial cells and monocytes in human subjects with asthma.
Frontiers in immunology, 12, 600979-600979 (2021-03-30)
Tenascin C (TNC) is an extracellular matrix glycoprotein that recently emerged as an immunomodulator. TNC-deficient (TNC-/-) mice were reported to have a reduced inflammatory response upon systemic administration of lipopolysaccharide, the toxic component of gram-negative bacteria. Here, we investigated the
The coordinated behavior of proteins is central to systems biology. However, the underlying mechanisms are poorly known and methods to analyze coordination by conventional quantitative proteomics are still lacking. We present the Systems Biology Triangle (SBT), a new algorithm that
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.