Skip to Content
Merck
All Photos(1)

Key Documents

AB1593

Sigma-Aldrich

Anti-Amyloid Precursor Protein Antibody, a.a. 44-63

serum, Chemicon®

Synonym(s):

APP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

goat

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... APP(351)

Specificity

Recognizes amyloid precursor protein (APP). In Alzheimer′s disease, this protein is broken down and releases the 12 amino acid b-amyloid protein that is found in senile plaques and vessels.

The amyloid precursor protein is the subject of intensive investigations to determine how this protein is broken down abnormally in Alzheimer′s disease to give rise to the b-amyloid protein which is present in senile plaques and vessels. This antiserum has been shown to be immunoreactive to the immunizing peptide by ELISA. The antiserum will immunolabel dystrophic neurites in senile plaques in both formalin-fixed paraffin sections and paraformaldehyde fixed frozen sections from Alzheimer′s brain.

This antiserum should be a valuable tool for scientist and neuropathologists who study the role of amyloid in Alzheimer′s′s disease.

Immunogen

Epitope: a.a. 44-63
Peptide to the N-terminal region (amino acids 44-63) of the APP protein.

Application

Anti-Amyloid Precursor Protein Antibody, a.a. 44-63 is an antibody against Amyloid Precursor Protein for use in ELISA, IF, WB, IH, IH(P).
Immunoblotting: 1:1,000

Immunofluorescence

Immunohistochemistry

Formalin fixed paraffin embedded tissue:1:50-1:100

4% paraformaldehyde fixed frozen sections: 1:2,500-1:5,000

ELISA: 1:1,500 against immunogen peptide

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Physical form

Goat antiserum. Liquid with 0.1% Sodium Azide

Storage and Stability

Store at -20ºC in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service