Skip to Content
Merck
All Photos(1)

Key Documents

AB1570

Sigma-Aldrich

Anti-GABA Transporter-1 Antibody, CT (a.a. 588-599)

Chemicon®, from rabbit

Synonym(s):

GAT-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

monkey, feline, rat, human, mouse, rabbit

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

suitability

not suitable for Western blot

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SLC6A1(6529)

Specificity

GABA Transporter-1 (GAT-1), C-terminus. No cross-reactivity to the C-termini of other transmitter transporters detected.

Immunogen

Epitope: C-terminus (a.a. 588-599)
Rat GAT-1, C-terminus (aa 588-599)

Application

Anti-GABA Transporter-1 Antibody, C-terminus (a.a. 588-599) is an antibody against GABA Transporter-1 for use in IH.
Immunohistochemistry: 1:500-1:750 on 4% paraformaldehyde fixed or 4% paraformaldehyde and 0.2% glutaraldehyde fixed tissue. It is recommended that the antibody be diluted in PBS containing 10% normal goat serum and 0.5% Triton X-100. AB1570 has been used for both light and electron microscopic studies of rat retina, cortex and hippocampus.

Note: AB1570 is not recommended for Western blot use. See AB1570W.

Optimal working dilutions must be determined by the end user.

Physical form

Affinity purified antibody. Lyophilized from in 0.1 M PBS with 1% BSA and 0.1% sodium azide. Reconstitute with 100 μL of sterile distilled water.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lu Pu et al.
Evidence-based complementary and alternative medicine : eCAM, 2012, 818451-818451 (2013-02-01)
Analgesia is a well-documented effect of acupuncture. A critical role in pain sensation plays the nervous system, including the GABAergic system and opioid receptor (OR) activation. Here we investigated regulation of GABA transporter GAT1 by δOR in rats and in
Muhamed N H Eeza et al.
Journal of Alzheimer's disease : JAD, 81(2), 797-808 (2021-04-13)
Circadian rhythm disturbance is commonly observed in Alzheimer's disease (AD). In mammals, these rhythms are orchestrated by the superchiasmatic nucleus (SCN). Our previous study in the Tg2576 AD mouse model suggests that inflammatory responses, most likely manifested by low GABA
Constantino Sotelo
The Journal of comparative neurology, 506(2), 240-262 (2007-11-21)
The acquisition of the dynamic balance between excitation and inhibition in developing Purkinje cells, necessary for their proper function, is analyzed. Newborn (P0) mouse cerebellum contains glutamatergic (VGLUT2-IR) and gamma-aminobutyric acid (GABA)-ergic (VIAAT-IR) axons. The former prevail and belong to
Pericellular innervation of neurons expressing abnormally hyperphosphorylated tau in the hippocampal formation of Alzheimer's disease patients.
Blazquez-Llorca, L; Garcia-Marin, V; Defelipe, J
Frontiers in Neuroanatomy null
Elena E Bagley et al.
Neuron, 45(3), 433-445 (2005-02-08)
Adaptations in neurons of the midbrain periaqueductal gray (PAG) induced by chronic morphine treatment mediate expression of many signs of opioid withdrawal. The abnormally elevated action potential rate of opioid-sensitive PAG neurons is a likely cellular mechanism for withdrawal expression.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service