Skip to Content
Merck
All Photos(1)

Key Documents

AB1431

Sigma-Aldrich

Anti-Interferon-β Antibody

serum, Chemicon®

Synonym(s):

IFN-beta

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
neutralization: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... IFNB1(3456)

General description

Interferon beta is a globular protein consisting of 5 alpha helices with a predicted molecular weight of 20 kDa. Due to glycosylation, however, the observed molecular weight is closer to 25 kDa by SDS-PAGE.

Specificity

Recognizes Human interferon beta.

Immunogen

Recombinant human interferon beta expressed in E. coli

Application

Interferon neutralization: 2 x 10e5 neutralization units/mL where one neutralization unit is the amount of antiserum required to neutralize one unit of human interferon beta (Hu-IFN-b) to a 50% endpoint. Interferon was titrated with the use of the cytopathic effect inhibition assay as described [Rubinstein, S., Familletti, P.C., and Pestka, S. (1981) "Convenient Assay for Interferons," in J. Virol., Vol. 37, 755-758]; [Familletti, P.C., Rubinstein, S., and Pestka, S. (1981) "A Convenient and Rapid Cytopathic Effect Inhibition Assay for Interferon," in Methods of Enzymology, Vol. 78 (S. Pestka, ed.), Academic Press, New York, 387-394]. In this antiviral assay for interferon about 1 unit/mL of interferon is the quantity necessary to produce a cytopathic effect of 50%. The units are determined with respect to the international reference standard for human interferon beta provided by the National Institutes of Health [see Pestka, S. (1986) "Interferon Standards and General Abbreviations," in Methods in Enzymology (S. Pestka, ed.), Academic Press, New York 119, 14-23]. This material is prepared specifically for the effective neutralization of human interferon beta.

Immunohistochemistry

Western blot: 1:5000

Immunoprecipitation

ELISA

Optimal working dilutions must be determined by end user.
Research Category
Inflammation & Immunology
Research Sub Category
Cytokines & Cytokine Receptors
This Anti-Interferon-β Antibody is validated for use in ELISA, IH, IP, NEUT, WB for the detection of Interferon-β.

Target description

20 kDa

Linkage

Replaces: 04-1095

Physical form

Rabbit Serum diluited with PBS. Liquid containing no preservatives.
Unpurified

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
IFN expressing cell lines

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Du et al.
Stem cells (Dayton, Ohio), 34(4), 960-971 (2015-12-18)
Fanconi anemia (FA) is an inherited bone marrow (BM) failure syndrome, presumably resulting from defects in hematopoietic stem cells (HSCs). Normal HSCs depend more on glycolysis than on oxidative phosphorylation (OXPHOS) for energy production. Here, we show that FA HSCs
Harumasa Nakazawa et al.
PloS one, 12(1), e0170391-e0170391 (2017-01-19)
Inflammation and apoptosis develop in skeletal muscle after major trauma, including burn injury, and play a pivotal role in insulin resistance and muscle wasting. We and others have shown that inducible nitric oxide synthase (iNOS), a major mediator of inflammation
Aisha Nazli et al.
Cellular & molecular immunology, 16(2), 178-194 (2018-03-20)
More than 40% of HIV infections occur via female reproductive tract (FRT) through heterosexual transmission. Epithelial cells that line the female genital mucosa are the first line of defense against HIV-1 and other sexually transmitted pathogens. These sentient cells recognize
Viacheslav Malikov et al.
Cell reports, 38(7), 110396-110396 (2022-02-17)
Fasciculation and elongation protein zeta-1 (FEZ1) is a multifunctional kinesin adaptor involved in processes ranging from neurodegeneration to retrovirus and polyomavirus infection. Here, we show that, although modulating FEZ1 expression also impacts infection by large DNA viruses in human microglia
Caroline L Ashley et al.
Viruses, 11(3) (2019-03-16)
The antiviral activity of type I interferons (IFNs) is primarily mediated by interferon-stimulated genes (ISGs). Induction of ISG transcription is achieved when type I IFNs bind to their cognate receptor and activate the Janus Kinase/Signal Transducer and Activator of Transcription

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service