Skip to Content
Merck
All Photos(1)

Key Documents

AB1258

Sigma-Aldrich

Anti-Cytochrome P450 Enzyme CYP1A1 Antibody

serum, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... AHRR(57491)

Specificity

Reacts with human cytochrome P450 CYP1A1 in human placental microsomal fraction and to recombinant human CYP1A1.

Immunogen

Synthetic peptide

Application

Anti-Cytochrome P450 Enzyme CYP1A1 Antibody detects level of Cytochrome P450 Enzyme CYP1A1 & has been published & validated for use in IH & WB.
Research Category
Metabolism
Research Sub Category
Enzymes & Biochemistry
Western blot

Immunohistochemistry

Optimal working dilutions must be determined by end user.

Linkage

Replaces: MAB3522

Physical form

Rabbit antiserum. Liquid, no preservatives.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Fei Yin et al.
Oncology reports, 30(1), 364-370 (2013-04-23)
Aryl hydrocarbon receptor (AhR) is a ligand-activated transcription factor associated with tumor initiation and progression. AhR expression is significantly increased in gastric cancer tissues and gastric cancer cell lines; however, the relationship between AhR and gastric cancer is still unclear.
A Kenneth MacLeod et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 24(9), 2138-2147 (2018-02-14)
Purpose: Osimertinib is a third-generation inhibitor of the epidermal growth factor receptor used in treatment of non-small cell lung cancer. A full understanding of its disposition and capacity for interaction with other medications will facilitate its effective use as a
Y Kapelyukh et al.
Drug metabolism and disposition: the biological fate of chemicals, 47(8), 907-918 (2019-05-31)
Cytochrome P450s CYP1A1 and CYP1A2 can metabolize a broad range of foreign compounds and drugs. However, these enzymes have significantly overlapping substrate specificities. To establish their relative contribution to drug metabolism in vivo, we used a combination of mice humanized
Elevated expression of CYP1A1 and gamma-SYNUCLEIN in human ectopic (ovarian) endometriosis compared with eutopic endometrium.
Singh, MN; Stringfellow, HF; Taylor, SE; Ashton, KM; Ahmad, M; Abdo, KR; El-Agnaf et al.
Molecular Human Reproduction null
D C Koch et al.
Oncogene, 34(50), 6092-6104 (2015-04-14)
The aryl hydrocarbon receptor (AhR) is a ligand-activated transcription factor and a member of the basic helix-loop-helix PER/ARNT/SIM family of chemosensors and developmental regulators. The AhR is widely known as a mediator of dioxin toxicity; however, it also suppresses cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service