Skip to Content
Merck
All Photos(1)

Key Documents

8.51221

Sigma-Aldrich

PyAOP

for peptide synthesis, Novabiochem®

Synonym(s):

PyAOP, (7-Azabenzotriazol-1-yloxy)trispyrrolidinophosphonium hexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H27F6N7OP2
CAS Number:
Molecular Weight:
521.38
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.22
Pricing and availability is not currently available.

Product Name

PyAOP, Novabiochem®

Quality Level

product line

Novabiochem®

form

powder

reaction suitability

reaction type: Coupling Reactions

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

15-25°C

SMILES string

F[P-](F)(F)(F)(F)F.[P](=[N+]5CCCC5)(N4CCCC4)(N3CCCC3)O[n]1nnc2c1nccc2

InChI

1S/C17H27N7OP.F6P/c1-2-11-21(10-1)26(22-12-3-4-13-22,23-14-5-6-15-23)25-24-17-16(19-20-24)8-7-9-18-17;1-7(2,3,4,5)6/h7-9H,1-6,10-15H2;/q+1;-1

InChI key

CBZAHNDHLWAZQC-UHFFFAOYSA-N

General description

PyAOP is an extremely effective coupling reagent that mediates amide bond formation with the efficiency of HATU but without the risk of guanidine formation, making it the reagent of choice for peptide cyclization.[1]

Associated Protocols and Technical Articles
Guide to Selection of Coupling Reagents

Literature references:
[1] F. Albericio, et al. (1997) Tetrahedron Lett., 38, 4853.

Application

PyAOP can be used as a coupling reagent:
  • In the synthesis of N,N′-substituted acylguanidines by solid-phase peptide synthesis.[2]
  • To facilitate the formation of peptide bonds during the synthesis of cyclic peptide hybrids.[3]

Analysis Note

Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Assay (HPLC, area%): ≥ 99.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of cyclic peptide hybrids with amino acid and nucleobase side-chains
Planas M, et al.
Tetrahedron Letters, 41, 4097-4100 (2000)
Solid-phase synthesis of N, N?-substituted acylguanidines
Dodd DS and Zhao Y
Tetrahedron Letters, 42, 1259-1262 (2001)
On the use of PyAOP, a phosphonium salt derived from HOAt, in solid-phase peptide synthesis
F. Albericio, et al.
Tetrahedron Letters, 38, 4853-4853 (1997)

Articles

Novabiochem® coupling reagents for in situ activation are fast-reacting and compatible with various amino acids.

Novabiochem® coupling reagents for in situ activation are fast-reacting and compatible with various amino acids.

Novabiochem® coupling reagents for in situ activation are fast-reacting and compatible with various amino acids.

Novabiochem® coupling reagents for in situ activation are fast-reacting and compatible with various amino acids.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service