Skip to Content
Merck
All Photos(1)

Key Documents

8.22256

Sigma-Aldrich

Aniline

for synthesis

Synonym(s):

Aniline, Aminobenzene, Phenylamine, Benzenamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7N
CAS Number:
Molecular Weight:
93.13
MDL number:
UNSPSC Code:
12352116
EC Index Number:
200-539-3
NACRES:
NA.22
Assay:
≥99.0% (GC)
bp:
184 °C/1013 hPa
vapor pressure:
0.5 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

0.5 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

540 °C

potency

840 mg/kg LD50, skin (Rabbit)

expl. lim.

1.2-11 % (v/v)

pH

8.8 (20 °C, 36 g/L in H2O)

bp

184 °C/1013 hPa

mp

-6.2 °C

transition temp

flash point 76 °C

solubility

36 g/L

density

1.02 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Nc1ccccc1

InChI

1S/C6H7N/c7-6-4-2-1-3-5-6/h1-5H,7H2

InChI key

PAYRUJLWNCNPSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • One pot conversion of phenols and anilines to aldehydes and ketones: Exploiting α gem boryl carbanions, this study offers a method compatible with a diverse range of phenols and anilines, facilitating simpler synthetic pathways in organic chemistry (Das et al., 2024).
  • In-silico-assisted derivatization of triarylboranes: This research discusses a catalytic method for the reductive alkylation of aniline-derived amino acids, improving functional-group compatibility in peptide modification (Hisata et al., 2024).
  • C–heteroatom coupling with electron-rich aryls: Utilizing nickel catalysis and light, this method enhances the synthesis of anilines and aryl ethers, contributing to more efficient organic synthesis processes (Cornella et al., 2024).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.020 - 1.022
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1 - STOT RE 1

Target Organs

Blood

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service