Skip to Content
Merck
All Photos(1)

Key Documents

8.21068

Sigma-Aldrich

Squalene

for synthesis

Synonym(s):

Squalene, 2,6,10,15,19,23-Hexamethyl-2,6,10,14,18,22-tetracosahexaene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H50
CAS Number:
Molecular Weight:
410.72
MDL number:
UNSPSC Code:
12352002
EC Index Number:
203-826-1
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥98.0% (GC)

form

liquid

kinematic viscosity

22.9 cSt(20 °C)

bp

335 °C/1013 hPa

transition temp

flash point >200 °C

density

0.86 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C(C\C=C(\CCC=C(C)C)/C)\C(=C\CC\C=C(\CC\C=C(\CCC=C(C)C)/C)/C)\C

InChI

1S/C30H50/c1-25(2)15-11-19-29(7)23-13-21-27(5)17-9-10-18-28(6)22-14-24-30(8)20-12-16-26(3)4/h15-18,23-24H,9-14,19-22H2,1-8H3/b27-17+,28-18+,29-23+,30-24+

InChI key

YYGNTYWPHWGJRM-AAJYLUCBSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.854 - 0.856
Peroxide value: ≤ 5
Identity (IR): passes test

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service