Skip to Content
Merck
All Photos(1)

Key Documents

8.20805

Sigma-Aldrich

Methyllithium

(5% solution in diethylether) for synthesis

Synonym(s):

Lithium methyl, MeLi

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
Pricing and availability is not currently available.

Quality Level

form

liquid

autoignition temp.

180 °C (Information refers to the main component.)

expl. lim.

1.7-36 % (v/v) Information refers to the main component.)

bp

34.6 °C (Information refers to the main component.)

transition temp

flash point -40 °C (Information refers to the main component.)

density

0.7 g/cm3 at 20 °C

storage temp.

2-8°C

General description

(5% solution in diethylether) for synthesis

Analysis Note

Assay (total alcalinity)4.5 - 5.5 %

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 1 - Pyr. Liq. 1 - Skin Corr. 1B - STOT SE 3 - Water-react. 1

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(C)

-40 °C - Information refers to the main component.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service