Skip to Content
Merck

8.20331

Methyl 2-chloropropionate

for synthesis

Synonym(s):

Methyl 2-chloropropionate, 2-Chloropropionic acid methyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C4H7ClO2
CAS Number:
Molecular Weight:
122.55
UNSPSC Code:
12352108
EC Index Number:
241-624-5
NACRES:
NA.22
MDL number:
Assay:
≥98.0% (GC)
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

30 hPa ( 20 °C)

Quality Level

assay

≥98.0% (GC)

form

liquid

autoignition temp.

430 °C (DIN 51794)

expl. lim.

2.8-13.9 % (v/v)

pH

6-7 (20 °C in H2O)

mp

<-10 °C

transition temp

flash point 36 °C (DIN 51755 Part 1)

density

1.13 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ClC(C)C(=O)OC

InChI

1S/C4H7ClO2/c1-3(5)4(6)7-2/h3H,1-2H3

InChI key

JLEJCNOTNLZCHQ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.136 - 1.139
Identity (IR): passes test


pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

100.4 °F - closed cup

flash_point_c

38 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
820331100004022536456614
820331025004022536456607