Skip to Content
Merck
All Photos(1)

Key Documents

8.18701

Sigma-Aldrich

Benzyl benzoate

for synthesis

Synonym(s):

Benzyl benzoate, Benzoic acid benzyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12O2
CAS Number:
Molecular Weight:
212.24
MDL number:
UNSPSC Code:
12352108
EC Index Number:
204-402-9
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

<0.1 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

480 °C

potency

1904 mg/kg LD50, oral (Rat)
4000 mg/kg LD50, skin (Rabbit)

bp

324 °C/1013 hPa

mp

21 °C

transition temp

flash point 158 °C

solubility

0.0153 g/L

density

1.12 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(Cc2ccccc2)C(=O)c1ccccc1

InChI

1S/C14H12O2/c15-14(13-9-5-2-6-10-13)16-11-12-7-3-1-4-8-12/h1-10H,11H2

InChI key

SESFRYSPDFLNCH-UHFFFAOYSA-N

Application

  • Essential oil therapy for inflammation: Research highlights the use of Cinnamomum zeylanicum essential oil, containing benzyl benzoate, in treating inflammation. Network pharmacology and experimental verification delineated the multitarget pharmacological mechanism, underscoring its potential as an anti-inflammatory agent (Mohanty et al., 2024).
  • Pest control in agriculture: Benzyl benzoate was evaluated for its effectiveness as an eco-friendly pesticide against the red flour beetle, Tribolium castaneum, demonstrating significant repellency and low toxicity, making it a viable option for sustainable pest management (Aboelhadid et al., 2023).
  • Scabies treatment: A comparative study on the efficacy of topical benzyl benzoate versus oral ivermectin for treating scabies revealed that benzyl benzoate is an effective topical treatment, providing a viable option for scabies management (Meyersburg et al., 2023).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.117 - 1.119
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

298.4 °F - closed cup

Flash Point(C)

148 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service