Skip to Content
Merck

8.17066

SAFC

Isopropyl myristate

EMPROVE® ESSENTIAL, Ph. Eur., NF

Pharma Manufacturing

Synonym(s):

Isopropyl myristate, Tetradecanoic acid isopropyl ester, Myristic acid isopropyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H34O2
CAS Number:
Molecular Weight:
270.45
MDL number:
UNSPSC Code:
12352108
EC Index Number:
203-751-4
Pricing and availability is not currently available.

Agency

NF
Ph. Eur.

Quality Level

vapor pressure

<1 hPa ( 20 °C)

product line

EMPROVE® ESSENTIAL

form

liquid

autoignition temp.

>300 °C

bp

140 °C/3 hPa

transition temp

flash point >150 °C

density

0.85 g/cm3 at 20 °C

application(s)

liquid formulation
pharmaceutical
semi-solid formulation

storage temp.

15-25°C

SMILES string

O(C(C)C)C(=O)CCCCCCCCCCCCC

InChI

1S/C17H34O2/c1-4-5-6-7-8-9-10-11-12-13-14-15-17(18)19-16(2)3/h16H,4-15H2,1-3H3

InChI key

AXISYYRBXTVTFY-UHFFFAOYSA-N

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Isopropyl myristate is used as solvent, penetration enhancer and emulsifying component in topical formulations, mainly semi-solid dosage forms.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

awg

Flash Point(F)

>302.0 °F

Flash Point(C)

> 150 °C


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Learn more about our culture media portfolio for microbial examination of non-sterile products, fully compliant with the harmonized Pharmacopoeia.

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service