Skip to Content
Merck

Skip To

8.09692

o-Cresol

for synthesis

Synonym(s):

o-Cresol, 2-Hydroxytoluene, 2-Methylphenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
2-(CH3)C6H4OH
CAS Number:
Molecular Weight:
108.14
UNSPSC Code:
12352104
EC Index Number:
202-423-8
NACRES:
NA.22
MDL number:
Assay:
≥99% (GC)
Grade:
synthesis grade
Bp:
191-192 °C/1013 hPa
Vapor pressure:
0.24 hPa ( 20 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

synthesis grade

Quality Segment

vapor pressure

0.24 hPa ( 20 °C)

assay

≥99% (GC)

form

solid

autoignition temp.

555 °C

potency

121 mg/kg LD50, oral (Rat), 890 mg/kg LD50, skin (Rabbit)

expl. lim.

1.3 % (v/v)

dilution

(for synthesis)

pH

4.8 (20 °C, 20 g/L in H2O)

bp

191-192 °C/1013 hPa

mp

31 °C

transition temp

flash point 81 °C

solubility

20 g/L

density

1.046 g/cm3 at 20 °C

storage temp.

no temp limit

SMILES string

Oc1c(cccc1)C

InChI

1S/C7H8O/c1-6-4-2-3-5-7(6)8/h2-5,8H,1H3

InChI key

QWVGKYWNOKOFNN-UHFFFAOYSA-N

Application

o-Cresol can be used as a starting material to synthesize:
  • Carvacrol by alkylation reaction with propylene or isopropyl alcohol in the presence of Zr/mesoporous silica as a catalyst.
  • Polyisobutylenes by selective cationic polymerization of isobutylene using AlCl3 as a catalyst.
  • Poly(o-cresol) via oxidative coupling polymerization reaction in the presence of substituted pyridine/CuCl catalysts.

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 29 °C
Melting range (upper value): ≤ 33 °C
Water (K. F.): ≤ 0.10 %
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
05-64708.052238.02295
grade

synthesis grade

grade

SAJ first grade

grade

synthesis grade

grade

-

Quality Level

200

Quality Level

-

Quality Level

200

Quality Level

200

assay

≥99% (GC)

assay

≥97.0%

assay

≥98.0% (GC)

assay

≥99.0% (GC)

solubility

20 g/L

solubility

-

solubility

-

solubility

3.9 g/L

form

solid

form

powder

form

solid

form

powder

vapor pressure

0.24 hPa ( 20 °C)

vapor pressure

0.3 mmHg ( 20 °C)

vapor pressure

0.06 hPa ( 20 °C)

vapor pressure

<0.1 hPa ( 20 °C)


Still not finding the right product?

Explore all of our products under o-Cresol


pictograms

Skull and crossbonesCorrosion

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 2

flash_point_f

177.8 °F - closed cup

flash_point_c

81.0 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Hello, Is it possible to get o-cresol in liqudi form?

    1 answer
    1. While the physical form of this product is not a specification, the material may present as a liquid or a solid. The melting point of this compound is approximately 31°C, gentle warming will re-liquify the product without harming the quality.
      There are other o-Cresol product options available that report the physical form on a lot-specific basis. This information is available in the product Certificate of Analysis. Please see the following links to review these products:
      https://www.sigmaaldrich.com/product/sigald/c85700
      https://www.sigmaaldrich.com/product/aldrich/w348007

      Please see the links below to review a sample or lot-specific Certificate for each item listed above. It is recommended to preview a Certificate of Analysis for current lots in stock, before purchasing either product:
      https://www.sigmaaldrich.com/product/sigald/c85700#product-documentation
      https://www.sigmaaldrich.com/product/aldrich/w348007#product-documentation

      Helpful?

Reviews

No rating value

Active Filters