Skip to Content
Merck
All Photos(1)

Key Documents

8.03598

Sigma-Aldrich

Divinylbenzene

(stabilised with 4-tert-butylpyrocatechol) for synthesis

Synonym(s):

Divinylbenzene, DVB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10
CAS Number:
Molecular Weight:
130.19
MDL number:
UNSPSC Code:
12352005
EC Index Number:
215-325-5
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

0.9 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

470 °C

potency

4100 mg/kg LD50, oral (Rat)
8000 mg/kg LD50, skin (Rabbit)

expl. lim.

1.1-6.2 % (v/v)

bp

195 °C/1013 hPa

mp

-88 °C

transition temp

flash point 77 °C

solubility

5 mg/L

density

0.91 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

c1(c(cccc1)C=C)C=C

InChI

1S/C10H10/c1-3-9-7-5-6-8-10(9)4-2/h3-8H,1-2H2

InChI key

MYRTYDVEIRVNKP-UHFFFAOYSA-N

General description

(stabilised with 4-tert-butylpyrocatechol) for synthesis

Application

Divinylbenzene (stabilised with 4-tert-butylpyrocatechol) for synthesis. CAS 1321-74-0, molar mass 130.18 g/mol.

Analysis Note

Assay isomers (GC, area%)60.0 - 65.0 % (a/a)Density (d 20 °C/ 4 °C)0.913 - 0.916Assay isomers ethylvinyl benzol (GC, area%)34.0 - 39.0 % (a/a)Identity (IR)passes testThe material is stabilised with 4-tert-Butylpyrocatechol. At time of analytical testing a content of >300 ppm stabiliser was determined.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

170.6 °F

Flash Point(C)

77 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service