Skip to Content
Merck

8.02485

Chromium(III) acetylacetonate

for synthesis

Synonym(s):

Chromium(III) acetylacetonate, Tris(acetylacetonato)chromium(III), Tris(2,4-pentanedionato)chromium(III)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H21CrO6
CAS Number:
Molecular Weight:
349.32
UNSPSC Code:
12352103
EC Index Number:
244-526-0
NACRES:
NA.22
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

form

powder

potency

3360 mg/kg LD50, oral (Rat), 6350 mg/kg LD50, skin (Rabbit)

pH

6 (20 °C, 1 g/L in H2O)

mp

213-216 °C

solubility

11 g/L

bulk density

450 kg/m3

storage temp.

2-30°C

SMILES string

[Cr](OC(=CC(=O)C)C)(OC(=CC(=O)C)C)OC(=CC(=O)C)C

InChI

1S/3C5H8O2.Cr/c3*1-4(6)3-5(2)7;/h3*3,6H,1-2H3;/q;;;+3/p-3

InChI key

JWORPXLMBPOPPU-UHFFFAOYSA-K

Analysis Note

Assay (iodometric): ≥ 98.0 %
Identity (IR): passes test


Still not finding the right product?

Explore all of our products under Chromium(III) acetylacetonate


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

392.0 °F

flash_point_c

> 200 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
802485999904022536373690
802485010004022536373676