Skip to Content
Merck
All Photos(1)

Key Documents

8.00429

Sigma-Aldrich

trans-Anethole

for synthesis

Synonym(s):

trans-Anethole, 1-(4-Methoxyphenyl)-1-propene, 4-Propenylanisole, 1-Methoxy-4-propenylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12O
CAS Number:
Molecular Weight:
148.20
MDL number:
UNSPSC Code:
12352112
EC Index Number:
224-052-0
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

1.33 hPa ( 63 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

potency

2090 mg/kg LD50, oral (Rat)
>5000 mg/kg LD50, skin (Rabbit)

pH

7 ( in H2O)

bp

235-237 °C/1013 hPa

mp

21-23 °C

transition temp

flash point 96 °C

density

0.99 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C)c1ccc(cc1)\C=C\C

InChI

1S/C10H12O/c1-3-4-9-5-7-10(11-2)8-6-9/h3-8H,1-2H3/b4-3+

InChI key

RUVINXPYWBROJD-ONEGZZNKSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.987 - 0.989
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

213.8 °F

Flash Point(C)

101 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service