Skip to Content
Merck
All Photos(1)

Key Documents

677175-M

Millipore

Vinblastine Sulfate

Antitumor alkaloid drug.

Synonym(s):

Vinblastine Sulfate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H58N4O9 · xH2SO4
CAS Number:
Molecular Weight:
810.97 (free base basis)
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

description

Merck USA index - 14, 9982

Assay

≥98% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

white

solubility

water: 50 mg/mL

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(O)O.N21C[C@H](C[C@@](C2)(O)CC)C[C@@](c8[nH]c9c(c8CC1)cccc9)(c3cc4c(cc3OC)N([C@@H]5[C@]64[C@H]7N(CC6)CC=C[C@]7([C@H]([C@]5(O)C(=O)OC)OC(=O)C)CC)C)C(=O)OC

InChI

1S/C46H58N4O9.H2O4S/c1-8-42(54)23-28-24-45(40(52)57-6,36-30(15-19-49(25-28)26-42)29-13-10-11-14-33(29)47-36)32-21-31-34(22-35(32)56-5)48(4)38-44(31)17-20-50-18-12-16-43(9-2,37(44)50)39(59-27(3)51)46(38,55)41(53)58-7;1-5(2,3)4/h10-14,16,21-22,28,37-39,47,54-55H,8-9,15,17-20,23-26H2,1-7H3;(H2,1,2,3,4)/t28-,37+,38-,39-,42+,43-,44-,45+,46+;/m1./s1

InChI key

KDQAABAKXDWYSZ-JKDPCDLQSA-N

General description

Antitumor alkaloid drug. Induces apoptosis in cultured hepatocytes and human lymphoma cells. Affects interaction of tubulin with microtubule-associated proteins, specifically Tau and MAP2. Depolymerizes microtubules. Increases the frequency of micronucleated erythrocytes.

Biochem/physiol Actions

Primary Target
Interaction of tubulin with microtubule-associated proteins, specifically Tau and MAP2

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Tanaka, E., et al. 1995. J. Cell. Biol.128, 139.
Boscoboinik, D., et al. 1994. Biochem. Biophys. Res. Commun.199, 374.
Smith, C.D., et al. 1994. Cancer Lett.79, 213.
Smith, C.D., et al. 1994. Cancer Res.54, 3779.
Tsukidate, K., et al. 1993. Am. J. Pathol. 143, 198.
Jagetia, G.C., and Jacob, P.S. 1992. Mutat. Res.280, 87.
Jordan, M.A., et al. 1992. J. Cell Sci. 102, 401.
Prasad, V., et al. 1992. J. Protein Chem.11, 509.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Muta. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service