Skip to Content
Merck
All Photos(1)

Key Documents

614310

Sigma-Aldrich

TLR3/dsRNA Complex Inhibitor

≥99% (HPLC), solid, TLR3/dsRNA complex inhibitor, Calbiochem®

Synonym(s):

TLR3/dsRNA Complex Inhibitor, ( R)-2-(3-Chloro-6-fluorobenzo[b]thiophene-2-carboxamido)-3-phenylpropanoic acid, Toll-Like Receptor 3/Double Strand RNA Complex Inhibitor, (R)-2-(3-Chloro-6-fluorobenzo[b]thiophene-2-carboxamido)-3-phenylpropanoic acid, Toll-Like Receptor 3/Double Strand RNA Complex Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H13ClFNO3S
Molecular Weight:
377.82
UNSPSC Code:
12352200
NACRES:
NA.28

product name

TLR3/dsRNA Complex Inhibitor, The TLR3/dsRNA Complex Inhibitor blocks dsRNA binding to TLR3.

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

shipped in

wet ice

storage temp.

2-8°C

General description

A thiophenecarboxamidopropionate compound that acts as a direct, competitive and high affinity inhibitor of dsRNA binding to TLR3 (Ki = 2.96 µM; Kd = 19 nM for dsRNA binding to TLR3) and selectively antagonizes stimulated TLR3 signaling without affecting TLR1/2, TLR2/6, TLR4 and TLR7 pathways. Shown to efficiently repress TNF-α and IL-1β (IC90 = 27 µM) and NO production (IC50 = 3.44 µM) in Poly(I:C)-induced RAW 264.7 macrophages. Exhibits minimal cytotoxicity (IC50 >>100 µM in RAW 264.7 cells) and is weakly active against a panel of 12-representative kinases and 4-cytochrome enzymes (in vitro assays).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Cheng, K., et al. 2011. J. Am. Chem. Soc.133, 3764.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer Maries Go Yap et al.
Frontiers in immunology, 13, 921728-921728 (2022-08-10)
Fibroblasts of different origins are known to possess stromal memory after inflammatory episodes. However, there are no studies exploring human lung fibroblast memory which may predict a subsequent inflammatory response in chronic respiratory diseases and COVID-19. MRC-5 and HF19 human
Tshifhiwa Magoro et al.
The Journal of biological chemistry, 294(40), 14591-14602 (2019-08-04)
Zika virus (ZIKV)3 is an enveloped, single-stranded, positive-sense RNA virus of the Flaviviridae family that has emerged as a public health threat because of its global transmission and link to microcephaly. Currently there is no vaccine for this virus. Conversion
Ki-Sun Kim et al.
Journal of leukocyte biology, 109(5), 865-875 (2021-02-23)
Although type I IFNs (IFN-I) are important for the innate and adaptive immune responses to suppress viral replication, prolonged IFN-I signaling in macrophages suppresses the immune response. Nuclear receptor estrogen-related receptor γ (ERRγ) regulates the transcription of genes involved in
Nina Pennetzdorfer et al.
Frontiers in cellular and infection microbiology, 13, 1250339-1250339 (2023-11-15)
Pseudomonas aeruginosa is a major human pathogen, particularly effective at colonizing the airways of patients with cystic fibrosis. Bacteriophages are highly abundant at infection sites, but their impact on mammalian immunity remains unclear. We previously showed that Pf4, a temperate
Lechuang Chen et al.
Scientific reports, 12(1), 21779-21779 (2022-12-17)
Elevated serum cytokine production in COVID-19 patients is associated with disease progression and severity. However, the stimuli that initiate cytokine production in patients remain to be fully revealed. Virus-infected cells release virus-associated exosomes, extracellular vesicles of endocytic origin, into the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service