Skip to Content
Merck
All Photos(1)

Key Documents

5.89592

Sigma-Aldrich

m-Xylene

Synonym(s):

m-Xylene, 1,3-Dimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
Beilstein:
605441
MDL number:
UNSPSC Code:
12191501
NACRES:
NA.22
grade:
anhydrous
Assay:
≥99%
bp:
138-139 °C (lit.)
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

vapor density

3.7 (vs air)

Assay

≥99%

autoignition temp.

982 °F

expl. lim.

7 %

refractive index

n20/D 1.497 (lit.)

bp

138-139 °C (lit.)

mp

−48 °C (lit.)

density

0.868 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(C)c1

InChI

1S/C8H10/c1-7-4-3-5-8(2)6-7/h3-6H,1-2H3

InChI key

IVSZLXZYQVIEFR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

m-Xylene may be used in the synthesis of 2,4-dimethylbenzophenone via Friedel–Craft acylation with benzoyl chloride over iron-modified tungstophosphoric acid supported on titania.citation It may also undergo chloromethylation in the presence of organic acids under organic-aqueous biphasic conditions to form a mixture of chloromethyl-2,4-dimethylbenzene and 1,3-bis(chloromethyl)-4,6-dimethylbenzene.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service