Skip to Content
Merck
All Photos(1)

Key Documents

566660

Sigma-Aldrich

SAG

≥98% (HPLC), solid, smoothened agonist, Calbiochem®

Synonym(s):

Smoothened Agonist, SAG

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H28ClN3OS · 2HCl · 2H2O
CAS Number:
Molecular Weight:
599.01
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

Smoothened Agonist, SAG, A cell-permeable Smoothened Agonist, SAG, CAS 364590-63-6, modulates the coupling of Smo with its downstream effector by interacting with the Smo heptahelical domain (KD = 59 nM).

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

light yellow

solubility

DMSO: 10 mg/mL
water: 25 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[s]1c2c(c(c1C(=O)N(C5CCC(CC5)NC)Cc3cc(ccc3)c4ccncc4)Cl)cccc2.Cl.Cl.O

InChI

1S/C28H28ClN3OS.2ClH.H2O/c1-30-22-9-11-23(12-10-22)32(28(33)27-26(29)24-7-2-3-8-25(24)34-27)18-19-5-4-6-21(17-19)20-13-15-31-16-14-20;;;/h2-8,13-17,22-23,30H,9-12,18H2,1H3;2*1H;1H2

InChI key

IYXUQUYRWPGIQL-UHFFFAOYSA-N

General description

A cell-permeable Smoothened Agonist, SAG, CAS 364590-63-6, modulates the coupling of Smo with its downstream effector by interacting with the Smo heptahelical domain (KD = 59 nM).
A cell-permeable benzothiophene compound that modulates the coupling of Smo with its downstream effector by interacting with the Smo heptahelical domain (KD = 59 nM). Shown to induce Smo internalization. Shown to induce Hedgehog pathway activation (EC50 ∼3 nM in NIH 3T3-derived Shh-LIGHT2 cells) and counteracts Cyclopamine-KAAD (Cat. No. 239804) inhibition of Smo. Reported to act as an activator at low concentrations and as an inhibitor at very high concentrations.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Wang, J., et al. 2010. Proc. Natl. Acad. Sci. USA.107, 9323.
Meloni, A.R., et al. 2006. Mol. Cell. Biol.26, 7750.
Chen, W., et al. 2004. Science306, 2257.
Chen, J.K., et al. 2002. Proc. Natl. Acad. Sci. USA99, 14071.
Frank-Kamenetsky, M., et al. 2002. J. Biol.1, 10.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ani Azatyan et al.
Cancers, 13(20) (2021-10-24)
Within the past decade, circular RNAs have largely emerged as novel regulators of human biology, including brain function and cancer development. On the other hand, the Hedgehog pathway has established roles in regulating biological processes, including tumorigenesis. Here, the circular
Denise Zujur et al.
Regenerative therapy, 14, 19-31 (2020-01-29)
Clinically relevant human induced pluripotent stem cell (hiPSC) derivatives require efficient protocols to differentiate hiPSCs into specific lineages. Here we developed a fully defined xeno-free strategy to direct hiPSCs toward osteoblasts within 21 days. The strategy successfully achieved the osteogenic
Zhourui Ma et al.
Journal of molecular histology, 51(3), 241-250 (2020-05-14)
Using a large-scale quantitative mesenchymal stem cells (MSCs) membrane proteomics analysis, we identified a large group of ciliary proteins in the MSCs membrane fraction, which prompted us to examine the cilia, intricate organelles that were originally discovered approximately 100 years ago.
Laura Girardet et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(6), 7593-7609 (2020-04-14)
Primary cilia (PC) are organelles that sense and respond to dynamic changes of the extracellular milieu through the regulation of target genes. By using the epididymis as a model system, we determined the contribution of primary cilia in the regulation
Shoko Onodera et al.
Stem cell reports, 15(1), 125-139 (2020-06-13)
Two genetic diseases, Gorlin syndrome and McCune-Albright syndrome (MAS), show completely opposite symptoms in terms of bone mineral density and hedgehog (Hh) activity. In this study, we utilized human induced pluripotent stem cell (iPSC)-based models of the two diseases to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service