Skip to Content
Merck
All Photos(1)

Key Documents

565771

Sigma-Aldrich

γ-Secretase Inhibitor X

≥90% (HPLC), liquid, γ-secretase inhibitor, Calbiochem®

Synonym(s):

InSolution γ-Secretase Inhibitor X, L-685,458, {1S-Benzyl-4R-[1-(1S-carbamoyl-2-phenethylcarbamoyl)-1S-3-methylbutylcarbamoyl]-2R-hydroxy-5-phenylpentyl}carbamic Acid tert-butyl Ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H52N4O6
Molecular Weight:
672.85
UNSPSC Code:
12352200
NACRES:
NA.54

product name

γ-Secretase Inhibitor X, InSolution, ≥90%, 1 mM

Quality Level

Assay

≥90% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

shipped in

ambient

storage temp.

−20°C

General description

A cell-permeable hydroxyethylene dipeptide isostere that acts as a highly specific and a potent inhibitor of γ-secretase (Aβtotal IC50 = 17 nM, Aβ40 IC50 = 48 nM, and Aβ42 IC50 = 67 nM in SHSY5Y cells overexpressing spβA4CTF). Binds to presenilin and blocks Notch intracellular domain production. Functions as a transition state analog mimic at the catalytic site of an aspartyl protease, however, it exhibits over 100-fold greater selectivity for γ-secretase than for cathepsin D.
A cell-permeable hydroxyethylene dipeptide isostere that acts as a potent and highly specific inhibitor of γ-secretase (Aβ total IC50 = 17 nM; Aβ40 IC50 = 48 nM; and Aβ42 IC50 = 67 nM in SH-SY5Y cells overexpressing spβA4CTF). Binds to presenilin and blocks Notch intracellular domain production. Also reported to block the formation of εCTF, resulting in the accumulation of α- and βCTF. Functions as a transition state analog mimic at the catalytic site of an aspartyl protease, however, it exhibits over 100-fold greater selectivity for γ-secretase than for cathepsin D.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
total
Product does not compete with ATP.
Reversible: no
Target IC50: Aβtotal 17 nM, Aβ40 48 nM, and Aβ42 67 nM in SHSY5Y cells overexpressing spβA4CTF

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

A 1 mM (250 µg in 372 µl or 500 µg in 744 µl) solution in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Weidemann, A., et al. 2002. Biochemistry41, 2825.
Doerfler, P., et al. 2001. Proc. Natl. Acad. Sci. USA98, 9312.
Li, Y.M., et al. 2000. Proc. Natl. Acad. Sci. USA97, 6138.
Shearman, M.S., et al. 2000. Biochemistry39, 8698.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Giulia Puliatti et al.
Progress in neurobiology, 227, 102482-102482 (2023-06-16)
Several studies including ours reported the detrimental effects of extracellular tau oligomers (ex-oTau) on glutamatergic synaptic transmission and plasticity. Astrocytes greatly internalize ex-oTau whose intracellular accumulation alters neuro/gliotransmitter handling thereby negatively affecting synaptic function. Both amyloid precursor protein (APP) and
Sylvia M Lee et al.
Cancer, 121(3), 432-440 (2014-09-25)
Aberrant Notch activation confers a proliferative advantage to many human tumors, including melanoma. This phase 2 trial assessed the antitumor activity of RO4929097, a gamma-secretase inhibitor of Notch signaling, with respect to the progression-free and overall survival of patients with
Lukas P Feilen et al.
eLife, 11 (2022-05-18)
Cleavage of membrane proteins in the lipid bilayer by intramembrane proteases is crucial for health and disease. Although different lipid environments can potently modulate their activity, how this is linked to their structural dynamics is unclear. Here, we show that
Nicole L Stott Bond et al.
Biomedicines, 11(1) (2023-01-22)
Lung cancer maintains a relatively small survival rate (~19%) over a 5-year period and up to 80-85% of all lung cancer diagnoses are Non-Small Cell Lung Cancer (NSCLC). To determine whether metformin reduces non-small cell lung cancer (NSCLC) LL/2 cell
Edgar Dawkins et al.
The Journal of biological chemistry, 299(4), 103027-103027 (2023-02-23)
Imbalances in the amounts of amyloid-β peptides (Aβ) generated by the membrane proteases β- and γ-secretase are considered as a trigger of Alzheimer's disease (AD). Cell-free studies of γ-secretase have shown that increasing membrane thickness modulates Aβ generation but it

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service