Skip to Content
Merck
All Photos(1)

Key Documents

557550

Sigma-Aldrich

Ro106-9920

A cell-permeable tetrazolopyridazine-phenylsulfoxide compound that displays anti-inflammatory properties.

Synonym(s):

Ro106-9920

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H7N5OS
CAS Number:
Molecular Weight:
245.26
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

white

solubility

ethanol: 1 mg/mL
DMSO: 25 mg/mL

shipped in

wet ice

storage temp.

−70°C

SMILES string

[S](=O)(c3ccccc3)c1n[n]2nnnc2cc1

InChI

1S/C10H7N5OS/c16-17(8-4-2-1-3-5-8)10-7-6-9-11-13-14-15(9)12-10/h1-7H

InChI key

JFSXSNSCPNFCDM-UHFFFAOYSA-N

General description

A cell-permeable tetrazolopyridazine-phenylsulfoxide compound that displays anti-inflammatory properties. Acts as a highly selective, irreversible inhibitor of IκBαee ubiquitination (IC50 = 2.3 µM). Also reported to prevent degradation of IκBαee in MM6 cells (complete inhibition at ~3-5 µM). Shown to block NF-κB-dependent cytokine expression in human peripheral blood mononuclear cells (PBMN) (IC50 = 600-700 nM for TNF-α, IL-1β, and IL-6 inhibition). Also shown to lower circulating levels of TNFα in LPS-treated rats. Also shown to inhibit 5-lipoxygenase (89% inhibition) and EGFR kinase (63% inhibition) at higher concentrations (10 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
IκBαee ubiquitination
Product does not compete with ATP.
Reversible: no
Target IC50: 2.3 µM against IκBαee ubiquitination

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-70°C). Stock solutions are stable for up to 3 months at -70°C.

Other Notes

Swinney, D.C., et al. 2002. J. Biol. Chem.277, 23573.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service