Skip to Content
Merck
All Photos(1)

Key Documents

540221

Sigma-Aldrich

Purine Nucleoside Phosphorylase, Human, Recombinant, E. coli

Synonym(s):

PNP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.42
Pricing and availability is not currently available.

recombinant

expressed in E. coli

Quality Level

form

liquid

specific activity

≥25 units/mg protein

manufacturer/tradename

Calbiochem®

shipped in

dry ice

storage temp.

−20°C

General description

Recombinant, human purine nucleotide phosphorylase expressed in E. coli without tags. Purine nucleoside phosphorylase catalyzes the phosphorlysis of N-ribosidic bonds of purine nucleosides and deoxynucleosides in a reversible reaction. Human PNP is a target for therapeutic T-cell immune response intervention. Useful in the detection of inorganic phosphate from biochemical reactions.
Recombinant, human purine nucleotide phosphorylase expressed in E. coli without tags. Purine nucleoside phosphorylase catalyzes the phosphorolysis of N-ribosidic bonds of purine nucleosides and deoxynucleosides. This reaction is reversible. The human enzyme is a target for therapeutic immune response intervention.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will convert 1 µmol of MESG and 1 µmol of Pi into 1 µmol of 2-amino-6-mercapto-7-methylpurine and 1 µmol of ribose phosphate in 1 min at 25°C, pH 7.6.

Physical form

In 50 mM Tris-HCl, pH 7.6.

Analysis Note

Single band by SDS-PAGE

Other Notes

Filgueira de Azevedo, W.Jr., et al. 2003. Biochem. Biophys. Res. Commun.309, 917.
Silva, R.G., et al. 2003. Protein Expr. Purif.27, 158.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roman S Iwasaki et al.
Nucleic acids research, 48(17), e101-e101 (2020-08-17)
Recent efforts in biological engineering have made detection of nucleic acids in samples more rapid, inexpensive and sensitive using CRISPR-based approaches. We expand one of these Cas13a-based methods to detect small molecules in a one-batch assay. Using SHERLOCK-based profiling of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service