Skip to Content
Merck
All Photos(1)

Key Documents

5.33639

Sigma-Aldrich

Pyruvate Dehydrogenase Kinase Inhibitor, AZD7545

Synonym(s):

Pyruvate Dehydrogenase Kinase Inhibitor, AZD7545, PDK Inhibitor, ( R)-4-(3-Chloro-4-(3,3,3-trifluoro-2-hydroxy-2-methylpropanamido)phenylsulfonyl)-N,N-dimethylbenzamide, PDh Kinase Inhibitor, PDhK Inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18ClF3N2O5S
CAS Number:
Molecular Weight:
478.87
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

SMILES string

FC(F)(F)[C@@](O)(C)C(=O)Nc1c(cc(cc1)[S](=O)(=O)c2ccc(cc2)C(=O)N(C)C)Cl

InChI

1S/C19H18ClF3N2O5S/c1-18(28,19(21,22)23)17(27)24-15-9-8-13(10-14(15)20)31(29,30)12-6-4-11(5-7-12)16(26)25(2)3/h4-10,28H,1-3H3,(H,24,27)/t18-/m1/s1

InChI key

DTDZLJHKVNTQGZ-GOSISDBHSA-N

General description

A cell-permeable, bioavailable trifluomethylpropanamide compound that acts as a potent, selective, reversible, non-ATP-competitive inhibitor of pyruvate dehydrogenase kinases (IC50 = 36.8 & 6.4 nM for PDhK1 & PDhK2, respectively). Inhibits PDhK3 at much higher concentrations (IC50 = 600 nM) and robustly stimulates PDhK4 activity (>10 nM). Also shown to increase pyruvate dehydrogenase (PDh) activity in liver and muscle, and potentiates pyruvate oxidation (EC50 = 105 nM in rat hepatocytes). Occupies lipoyl domain-binding site of PDhK2 (Kd = 19.2 nM) and disrupts the association of PDhK2 and inner lipoyl-bearing domain 2 of dihydrolipoyl acetyltransferase. Improves 24-h glucose profile and eliminates postparandial elevation in blood glucose in Zucker (fa/Fa) rats (10 mg/kg, b.i.d, p.o.,7 days).

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
Pyruvate Dehydrogenase Kinase Inhibitor, AZD7545, CAS 252017-04-2, is a potent, selective, reversible inhibitor of PDHK2 (IC₅₀ = 6.4 nM). Inhibits PDHK1 at higher levels (IC₅₀ = 36.8 nM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
pyruvate dehydrogenase kinases
Target IC50: 36.8 & 6.4 nM for PDhK1 & PDhK2, respectively

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Li, J., et al. 2009. J. Biol. Chem.284, 34458.
Tuganova, A., et al. 2007. Biochemistry46, 8592.
Morrell, J.A., et al. 2003. Biochem. Soc. Trans.31, 1168.
Mayers, R.M., et al. 2003. Biochem. Soc. Trans.31, 1165.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service