Skip to Content
Merck
All Photos(1)

Key Documents

5.05275

Sigma-Aldrich

ICI 118551 Hydrochloride

Synonym(s):

ICI 118551 Hydrochloride, (±)-1-[2,3-(Dihydro-7-methyl-1H-inden-4-yl)oxy]-3-[(1-methylethyl)amino]-2-butanol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H27NO2 · xHCl
CAS Number:
Molecular Weight:
277.40 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

storage temp.

10-30°C

SMILES string

N([C@H]([C@@H](O)COc1c2c(c(cc1)C)CCC2)C)C(C)C

InChI

1S/C17H27NO2/c1-11(2)18-13(4)16(19)10-20-17-9-8-12(3)14-6-5-7-15(14)17/h8-9,11,13,16,18-19H,5-7,10H2,1-4H3/t13-,16-/m0/s1

InChI key

VFIDUCMKNJIJTO-BBRMVZONSA-N

General description

ICI-118,551 reduces hemangioblastoma viability by promoting cell apoptosis and inhibits the formation of hemangiospheres from VHL tumor cells. This antiangiogenic agent inhibits HIF-target genes. It also blocks constitutively activated HIF signaling in hemangioblastomas associated with von Hippel-Lindau disease. ICI-118,551 is a potent inhibitor of isoprenaline-stimulated adenylate cyclase activity in rabbit ciliary process cell preparations.

Biochem/physiol Actions

Primary Target
→2 adrenergic receptors
Reversible: yes
Target Ki: 1.2, 120 and 257 nM for →2, →1 and →3 receptors respectively.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Bilski, A., et al. 1983. J. Cardiovasc. Pharmacol.5, 430.
Cook, S. et al. 1993. Br. J .Pharmacol.109, 1140.
Ten Berge, R., et al. 1995. Eur. J. Pharmacol.275, 199.
Strosberg, A., et al. 1996. TiPS17, 373.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The beta2-adrenergic receptor antagonist ICI-118,551 blocks the constitutively activated HIF signalling in hemangioblastomas from von Hippel-Lindau disease
Cuesta AM, et al.
Scientific reports, 9(1), 10062-10062 (2019)
The beta2-adrenergic receptor antagonist ICI-118,551 blocks the constitutively activated HIF signalling in hemangioblastomas from von Hippel-Lindau disease
Cuesta AM, et al.
Scientific Reports, 9(1), 10062-10062 (2019)
ICI 118,551: an effective ocular hypotensive agent with selectivity for the ciliary process beta 2-adrenoceptor and with minimal cardiac side effects.
Nathanson JA
British Journal of Pharmacology, 83(3), 821-821 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service