Skip to Content
Merck
All Photos(1)

Key Documents

5.04820

Sigma-Aldrich

Cinnabarinic acid

Synonym(s):

Cinnabarinic acid, 2-Amino-3-oxo-3H-phenoxazine-1,9-di­carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H8N2O6
CAS Number:
Molecular Weight:
300.22
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

red

solubility

DMSO: 25 mM

storage temp.

−20°C

InChI

1S/C14H8N2O6/c15-10-6(17)4-8-12(9(10)14(20)21)16-11-5(13(18)19)2-1-3-7(11)22-8/h1-4H,15H2,(H,18,19)(H,20,21)

InChI key

FSBKJYLVDRVPTK-UHFFFAOYSA-N

General description

A partial, low-potency agonist highly selective for mGlu4 receptors (EC50 >100 µM, no effect on other mGlu subtype receptors). Used in assessing the functional roles of mGlu4 receptors in a variety of research areas, such as excitotoxic cell death, apoptosis, neuroinflammation, inflammation, and neuroimmune communication.

Biochem/physiol Actions

Primary Target
mGlu₄ receptors

Warning

Toxicity: Standard Handling (A)

Preparation Note

Slight warming is required for complete solubilization.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Fazio, Francesco, et al. 2012. Mol. Pharmacol.81, 643.

Volpi, C., et al. 2012. Neuropharmacol.63, 501.
Hiramatsu, R., et al. 2008. J. Cell Biochem.103, 42.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service