Skip to Content
Merck
All Photos(1)

Key Documents

459620

Sigma-Aldrich

Okadaic Acid sodium salt

≥98% (TLC), solid,film (colorless), protein phosphatase 2A and protein phosphatase 1 inhibitor, Calbiochem®

Synonym(s):

Okadaic Acid, Sodium Salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H67O13 · Na
Molecular Weight:
826.98
UNSPSC Code:
12352106
NACRES:
NA.77

Product Name

Okadaic Acid, Sodium Salt, Water-soluble analog of Okadaic Acid. Inhibits protein phosphatases 1 and 2A.

Quality Level

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to off-white

solubility

water: 1 mg/mL
DMSO: 20 mg/mL
ethanol: 20 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

Water-soluble analog of Okadaic Acid (Cat. No. 495604). Inhibits protein phosphatases 1 and 2A.

Biochem/physiol Actions

Cell permeable: no
Product does not compete with ATP.
Reversible: no
Target IC50: 10-15 nM, 0.1 nM against protein phosphatase 1, protein phosphatase 2A, respectively

Warning

Toxicity: Toxic (F)

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Gjertsen, B.T., et al. 1994. J. Cell Sci.107, 3363.
Kiguchi, K., et al. 1994. Cell Growth Differentiation5, 995.
Ohaka, Y., et al. 1993. Biochem. Biophys. Res. Commun.197, 916.
Gopalakrishna, R., et al. 1992. Biochem. Biophys. Res. Commun.189, 950.
Kreienbuhl, P., et al. 1992. Blood80, 2911.
Normura, M., et al. 1992. Biochemistry31, 11915.
Song, Q., et al. 1992. J. Cell Physiol.153, 550.
Tada, Y., et al. 1992. Immunopharmacol.24, 17.
Cohen, P., et al. 1990. Trends Biochem. Sci.15, 98.
Cohen, P. 1989. Annu. Rev. Biochem.58, 453.
Cohen, P., and Cohen, P.T.W. 1989. J. Biol. Chem.254 21435.
Haystead, T.A. et al. 1989. Nature337, 78.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protein phosphatases come of age.
P Cohen et al.
The Journal of biological chemistry, 264(36), 21435-21438 (1989-12-25)
Y Tada et al.
Immunopharmacology, 24(1), 17-24 (1992-07-01)
Okadaic acid is a potent tumor promoter and an inhibitor of serine/threonine-specific protein phosphatases. We studied the effect of okadaic acid in human T cell activation and phosphorylation of internal substrates. Okadaic acid at up to 4 nM enhanced phorbol
Q Song et al.
Journal of cellular physiology, 153(3), 550-556 (1992-12-01)
Gamma-radiation, tetrandrine, bistratene A, and cisplatin were all found to induce pronounced morphological changes characteristic of apoptosis and extensive DNA fragmentation in the human BM13674 cell line 8 h after treatment. Apoptosis induced in BM13674 cells by these diverse agents
P Kreienbühl et al.
Blood, 80(11), 2911-2919 (1992-12-01)
The phosphatase inhibitors okadaic acid and calyculin A were found to elicit or to modify several neutrophil responses, suggesting that dephosphorylation plays a regulatory role. The concentrations of okadaic acid (> or = 1 mumol/L) that were effective on neutrophil
M Nomura et al.
Biochemistry, 31(47), 11915-11920 (1992-12-01)
Smooth muscle myosin light chain kinase is phosphorylated at two sites (A and B) by different protein kinases. Phosphorylation at site A increases the concentration of Ca2+/calmodulin required for kinase activation. Diphosphorylated myosin light chain kinase was used to determine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service