Skip to Content
Merck
All Photos(1)

Documents

36-011

Sigma-Aldrich

Anti-nitro-α/β-Synuclein Antibody, clone nSyn12

clone nSyn12, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

nSyn12, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

human ... SNCA(6622)

Specificity

α-Synuclein nitrated at Tyr125 and Tyr136
β-Synuclein nitrated at Tyr130

Application

Anti-nitro-α/β-Synuclein Antibody, clone nSyn12 is an antibody against nitro-α/β-Synuclein for use in WB, IH.

Quality

rountinely evaluated on recombinant protein nitrated in vitro

Target description

α-Synuclein, Mr 14.5kDa
β-Synuclein, Mr 17kDa

Physical form

Format: Unpurified

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bregje W M de Wildt et al.
Biotechnology and bioengineering, 120(7), 2013-2026 (2023-05-06)
The transition in the field of bone tissue engineering from bone regeneration to in vitro models has come with the challenge of recreating a dense and anisotropic bone-like extracellular matrix (ECM). Although the mechanism by which bone ECM gains its
B I Giasson et al.
The Journal of biological chemistry, 274(12), 7619-7622 (1999-03-13)
alpha-Synuclein is a soluble presynaptic protein which is pathologically redistributed within intracellular lesions characteristic of several neurodegenerative diseases. Here we demonstrate that wild type and two mutant forms of alpha-synuclein linked to familial Parkinson's disease (Ala30 --> Pro and Ala53
J M Souza et al.
The Journal of biological chemistry, 275(24), 18344-18349 (2000-04-05)
Intracellular proteinaceous aggregates are hallmarks of many common neurodegenerative disorders, and recent studies have shown that alpha-synuclein is a major component of several pathological intracellular inclusions, including Lewy bodies in Parkinson's disease (PD) and glial cell inclusions in multiple system
Nicole Willems et al.
BMC veterinary research, 12, 10-10 (2016-01-14)
Intervertebral disc (IVD) disease is a common spinal disorder in dogs and degeneration and inflammation are significant components of the pathological cascade. Only limited studies have studied the cytokine and chemokine profiles in IVD degeneration in dogs, and mainly focused
Emilio Fernández-Espejo et al.
Antioxidants (Basel, Switzerland), 10(5) (2021-06-03)
Background. Salivary α-synuclein (aSyn) and its nitrated form, or 3-nitrotyrosine-α-synuclein (3-NT-αSyn), hold promise as biomarkers for idiopathic Parkinson's disease (IPD). Nitrative stress that is characterized by an excess of 3-nitrotyrosine proteins (3-NT-proteins) has been proposed as a pathogenic mechanism in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service