Skip to Content
Merck
All Photos(1)

Key Documents

3301

Sigma-Aldrich

17β-Estradiol

17β-Estradiol

≥97% (HPLC), solid, estrogenic hormone, Calbiochem

Synonym(s):

17β-Estradiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H24O2
CAS Number:
Molecular Weight:
272.38
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

17β-Estradiol, Most potent mammalian estrogenic hormone.

Quality Level

description

RTECS - KG2975000
Merck USA index - 14, 3703

Assay

≥97% (by assay)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white to off-white

solubility

ethanol: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

O[C@@H]1[C@@]2([C@H]([C@H]3[C@H](CC2)c4c(cc(cc4)O)CC3)CC1)C

InChI

1S/C18H24O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h3,5,10,14-17,19-20H,2,4,6-9H2,1H3/t14-,15-,16+,17+,18+/m1/s1

InChI key

VOXZDWNPVJITMN-ZBRFXRBCSA-N

Warning

Toxicity: Harmful & Carcinogenic / Teratogenic (E)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

Active Filters

  1. Bucharest
    • Reviews 2
    • Votes 2
    4 out of 5 stars.

    fit for the purpose

    Low solubility in water, as known. Would recommend the Cyclodextrin-encapsulated form (E4389) for application that require only water or media as solvent.

    Helpful?

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service