Skip to Content
Merck
All Photos(1)

Key Documents

324461

Sigma-Aldrich

E2F Inhibitor, HLM006474

The E2F Inhibitor, HLM006474, also referenced under CAS 353519-63-8, controls the biological activity of E2F. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

E2F Inhibitor, HLM006474, 7-((4-Ethoxy-3-methylphenyl)(2-pyridylamino)methyl)-2-methyl-quinolinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H25N3O2
CAS Number:
Molecular Weight:
399.48
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.28

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

N(C(c4cc(c(cc4)OCC)C)c2c(c3nc(ccc3cc2)C)O)c1ncccc1

InChI

1S/C25H25N3O2/c1-4-30-21-13-11-19(15-16(21)2)23(28-22-7-5-6-14-26-22)20-12-10-18-9-8-17(3)27-24(18)25(20)29/h5-15,23,29H,4H2,1-3H3,(H,26,28)

InChI key

CYNZBLNMIJNBSF-UHFFFAOYSA-N

General description

A cell-permeable 8-hydroxyquinoline compound that is reported to inhibit nuclear E2F complexes DNA binding activity (IC50 = 30 µM) and downregulate E2F expression in the melanoma cell line A375, resulting in eventual apoptosis induction without upregulating p53 expression as commonly seen with traditional chemotherapeutic agents, such as Cisplatin (Cat. No. 232120), doxorubicin (Cat. No. 324380), and VP-16 (Cat. No. 341205).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Ma, Y., et al. 2008. Cancer Res.68, 6292.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service