Skip to Content
Merck
All Photos(1)

Key Documents

317700

Sigma-Aldrich

Dkk-1 Inhibitor, WAY-262611, HCl

The Dkk-1 Inhibitor, WAY-262611 controls the biological activity of Dkk-1.

Synonym(s):

Dkk-1 Inhibitor, WAY-262611, HCl, (1-(4-(Naphthalen-2-yl)pyrimidin-2-yl)piperidin-4-yl)methanamine, Wnt Pathway Activator VI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H22N4 · xHCl
Molecular Weight:
318.42 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

yellow

solubility

DMSO: 25 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A 2-aminopyrimidine compound that counteracts the antagonizing effect of Dkk-1 (Dickkopf-1) against Wnt-3a/LRP5 interaction and prevents Dkk-1-mediated Kr2/LRP5/Dkk-1 complex formation and internalization. Shown to successfully rescue cellular response to Wnt-3a stimulation in the presence of Dkk-1 (EC50 = 0.63 µM in a cell-based TCF-luciferase assay) in vitro and double the tibia trabecular bone formation in OVX rats in a period of 28 days via an oral dose of 10 mg/kg/day in vivo.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Other Notes

Pelletier, J.C., et al. 2009. J. Med. Chem.52, 6962.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service