Skip to Content
Merck
All Photos(1)

Key Documents

2905-OP

Sigma-Aldrich

Bovine Serum Albumin

OmniPur® Grade, >= 99%, Fraction V, Cold Alcohol Isolation

Synonym(s):

OmniPur® BSA, Fraction V, Cold Alcohol Isolation, Albumin, Bovine Serum, Bovine Serum Albumin, Fraction V

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25
Pricing and availability is not currently available.

Quality Level

Assay

≥99±% (electrophoresis)

form

lyophilized powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

off-white

shipped in

wet ice

General description

Bovine serum albumin (BSA) is a non-glycosylated globular protein. This α-helical protein belongs to the serum albumin family.[1][2] BSA comprises three domains with two sub-domains each and has 17-disulfide bonds linking them.[1][2]

Application

OmniPur® BSA, Fraction V, Cold Alcohol Isolation has been used:
  • as a blocking agent in immunofluorescence assay[3]
  • as a component of wash buffer in flow cytometric analysis[4]
  • as a component of anti-clumping buffer in ClpXP Dumbbell assay, motility assay and wet preparation to remove unbound beads and unbound substrate-DNA-biotin[5]

Biochem/physiol Actions

Bovine Serum Albumin (BSA) is a crucial component for cell culture media, which can stimulate embryonic stem cells (hESCs) differentiation.[6] It helps in the transportation of drugs, hormones, and fatty acids.[1]

Warning

Toxicity: Standard Handling (A)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon P Hood et al.
eLife, 9 (2020-07-28)
We demonstrate that prostate cancer can be identified by flow cytometric profiling of blood immune cell subsets. Herein, we profiled natural killer (NK) cell subsets in the blood of 72 asymptomatic men with Prostate-Specific Antigen (PSA) levels < 20 ng
Juan Carlos Cordova et al.
Methods in molecular biology (Clifton, N.J.), 1486, 317-341 (2016-11-16)
Energy-dependent protein degradation is studied through the dual bead ClpXP motility assay. Processing of folded proteins involves recognition, unfolding, translocation, and degradation stages. A dual optical trap, in a passive force-clamp geometry, exhibits bead-to-bead displacements that directly follow subprocesses underlying
Yaxin Li et al.
Journal of medicinal chemistry, 64(3), 1570-1583 (2021-02-02)
Androgen receptor (AR) contributes to the progression of glioblastoma (GBM), and antiandrogen agents have the potential to be used for the treatment of GBM. However, AR mutation commonly happens in GBM, which makes the antiandrogen agents less effective. Heat shock
Geoffrey L Francis
Cytotechnology, 62(1), 1-16 (2010-04-08)
Albumin has a long historical involvement in design of media for the successful culture of mammalian cells, in both the research and commercial fields. The potential application of albumins, bovine or human serum albumin, for cell culture is a by-product
Tamara Topală et al.
Clujul medical (1957), 87(4), 215-219 (2014-01-01)
The continuous search for new molecules with therapeutic abilities has led to the synthesis and characterization of a large number of metal complexes, proven to exhibit potential as pharmacological agents through their antibacterial, antiviral, antifungal and antineoplastic properties. As serum

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service