Skip to Content
Merck
All Photos(1)

Key Documents

281751

Sigma-Aldrich

DAB, Tetrahydrochloride, 50X Concentrate

This DAB, Tetrahydrochloride, 50X Concentrate is validated for use in Immunoblotting, Immunohistochemistry.

Synonym(s):

DAB, Tetrahydrochloride, 50X Concentrate, 3,3ʹ-Diaminobenzidine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116133
NACRES:
NA.41
Pricing and availability is not currently available.

description

RTECS - DV8753000

Quality Level

form

solution

does not contain

preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

brown to dark brown

shipped in

ambient

storage temp.

2-8°C

SMILES string

[Cl-].[Cl-].[Cl-].[Cl-].Nc1c(ccc(c1)c2cc(c(cc2)N)N)N.[H+].[H+].[H+].[H+]

InChI

1S/C12H14N4.4ClH/c13-9-3-1-7(5-11(9)15)8-2-4-10(14)12(16)6-8;;;;/h1-6H,13-16H2;4*1H

InChI key

KJDSORYAHBAGPP-UHFFFAOYSA-N

Warning

Toxicity: Toxic (F)

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service