Skip to Content
Merck
All Photos(1)

Key Documents

252790

Sigma-Aldrich

DZNep

≥97% (HPLC), solid, EZH2 inhibitor, Calbiochem®

Synonym(s):

Histone Methyltransferase EZH2 Inhibitor, DZNep, 3-deazaneplanocin A, HCl; (1S,2R,5R)-5-(4-amino-1H-imidazo[4,5-c]pyridin-1-yl)-3-(hydroxymethyl)cyclopent-3-ene-1,2-diol, HCl, HMTase Inhibitor X

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N4O3 · xHCl · yH2O
CAS Number:
Molecular Weight:
262.26 (anhydrous free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Histone Methyltransferase EZH2 Inhibitor, DZNep, Histone Methyltransferase EZH2 Inhibitor, DZNep, CAS 120964-45-6, is a cell-permeable inhibitor of EZH2-mediated trimethylation of K27 on histone H3.

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

tan

solubility

DMSO: 10 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

Cl.[n]2(c3c(nc2)c(ncc3)N)[C@H]1[C@@H]([C@@H](C(=C1)CO)O)O

InChI

1S/C12H14N4O3.ClH/c13-12-9-7(1-2-14-12)16(5-15-9)8-3-6(4-17)10(18)11(8)19;/h1-3,5,8,10-11,17-19H,4H2,(H2,13,14);1H/t8-,10-,11+;/m1./s1

InChI key

UNSKMHKAFPRFTI-FDKLLANESA-N

General description

A cell-permeable compound that is shown (at 1 µM concentrations) to inhibit EZH2-mediated trimethylation of K27 on histone H3 and induces the expression of cell-cycle regulatory genes, p21 and p27, as well as the cell death regulator, FBXO32, in OCI-AML3 and HL-60 cells, whereby treatment with inhibitor increases p16 levels in the former, but not the latter of the two cultures. At concentrations between 200 nm and 2000 nM, this compound is found to dose-dependently deplete the expression of polycomb group proteins EZH2, SUZ12, and EED in cultured and primary AML cell extracts. At concentrations up to ≥ 1000 nM, DZNep dose-dependently increases the percentage of apoptotic cells up to > ~ 38%, with greater potency against OCI-AML3 than HL-60 cultures and inhibits colony growth up to > ~ 85% for both cell lines. In OCI-AML3 cultures, 1000 nM of treatment demonstrates a significant increase in the accumulation of cells in the G0/G1 phase (58.5%) with a concomitant decrease in the number of cells in S phase (35.2%) and G2/M phases (6.3%) of the cell cycle. When co-treated with panobinostat (PS), 200 nM to 1000nM DZNep is shown to decrease cell viability in OCI-AML3 and HL-60 cultures more effectively than DZNep alone, in a dose-dependent manner. DZNep (1mg/kg, twice per week, i.p.) and PS significantly prolong the survival of mice implanted with HL-60 cells compared to treatment with either compound alone. DZNep is also a known S-adenosyl-L-homocysteine (AdoHcy) hydrolase inhibitor. Also available as a 25 mM solution in H2O (Cat. No. 506069).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Physical form

Supplied as a hydrochloride salt.

Reconstitution

Following reconstitution, aliquot and freeze 9-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Fiscus, W., et al. 2009. Blood13, 2733.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoxia Su et al.
Journal of cellular and molecular medicine, 26(2), 375-384 (2021-12-08)
Oxidative stress, a hallmark of ageing, inhibits the osteogenic differentiation of bone marrow-derived mesenchymal stem cells in long bone. The dysfunction of the cellular antioxidant defence system is a critical cause of oxidative stress, but the mechanism of the decline

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service