Skip to Content
Merck
All Photos(1)

Key Documents

251760

Sigma-Aldrich

DAR-4M

A rhodamine-based photo-stable nitric oxide (NO) fluorescent indicator with a detection limit of ~10 nM.

Synonym(s):

DAR-4M, Diaminorhodamine-4M

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H26N4O3
Molecular Weight:
430.50
UNSPSC Code:
12352202
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

solution

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

red-violet

fluorescence

λex ~560 nm
λem ~575 nm

shipped in

ambient

storage temp.

−20°C

General description

A rhodamine-based photo-stable nitric oxide (NO) fluorescent indicator with a detection limit of ~10 nM. Reacts with NO, in the presence of O2, resulting in a triazolo-rhodamine analog (DAR-4M T) that exhibits about 840-fold greater fluorescence quantum efficiency. The fluorescence intensity is not dependent on pH, unlike DAF-2 (Cat. No. 251500).

Biochem/physiol Actions

Cell permeable: no
Primary Target
Nitric oxide (NO) fluorescent indicator
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Irritant (B)

Physical form

In DMSO.

Reconstitution

Following initial thaw, aliquot and freeze at (-20°C). Use immediately following dilution and discard any unused portion.

Other Notes

Kojima, H., et al. 2001. Anal. Chem.73, 1967.
Kojima, H., et al. 2000. Tetrahedron Lett.41, 69.

Legal Information

Not available for sale in Japan.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service