Skip to Content
Merck

219331

β-Catenin/Tcf Inhibitor II, PKF118-310

The β-Catenin/Tcf Inhibitor II, PKF118-310 controls the biological activity of β-Catenin/Tcf. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

β-Catenin/Tcf Inhibitor II, PKF118-310, 1,6-Dimethylpyrimido[5,6-e][1,2,4]triazine-5,7-dione, Wnt Pathway Inhibitor X

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C7H7N5O2
Molecular Weight:
193.16
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥95% (HPLC)
Form:
solid
Quality level:
Storage condition:
OK to freeze, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, protect from light

color

dark yellow

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N1(N=CN=C2C1=NC(=O)N(C2=O)C)C

InChI

1S/C7H7N5O2/c1-11-6(13)4-5(10-7(11)14)12(2)9-3-8-4/h3H,1-2H3

InChI key

SLGRAIAQIAUZAQ-UHFFFAOYSA-N

General description

A synthetic cell-permeable actinobacteria pyrimidotriazine-dione that selectively disrupts β-catenin-Tcf4 interaction (IC50 = 0.8 µM in cell-free binding assays) and inhibits β-catenin-regulated cellular transcription activity (IC50 = 0.3 µM in HCT116-based reporter assays). PKF118-310 is demonstrated to reverse β-catenin, but not siamois, mRNA-induced ventral body axis formation in Xenopus embryos, while direct tumor site injection (1 mg/kg/0.5 wk) in HepG2 xenograph mice is shown to increase the number of apoptotic cells and decrease the expression of β-catenin target genes (cyclin D1, c-Myc, survivin) in the tumor mass, resulting in an effective tumor growth retardation (by ~60% after a 4-wk treatment period) in vivo.

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Wei, W., et al. 2010. Int. J. Cancer126, 2426.
Lepourcelet, M., et al. 2004. Cancer Cell5, 91.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 2 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Yinfei Tan et al.
Scientific reports, 10(1), 15837-15837 (2020-09-29)
The Dlx5 homeobox gene was first implicated as an oncogene in a T-ALL mouse model expressing myristoylated (Myr) Akt2. Furthermore, overexpression of Dlx5 was sufficient to drive T-ALL in mice by directly activating Akt and Notch signaling. These findings implied



Global Trade Item Number

SKUGTIN
219331-10MG04055977202106