Skip to Content
Merck
All Photos(1)

Key Documents

217503

Sigma-Aldrich

Cardiotoxin

from Naja pallida, ≥95% (HPLC), lyophilized solid, cytolytic toxin, Calbiochem®

Synonym(s):

Cardiotoxin, Naja pallida

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

Product Name

Cardiotoxin, Naja pallida,

Assay

≥95% (HPLC)

Quality Level

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

2-8°C

General description

Cardiotoxins, which are highly basic peptides, interact with negatively charged lipids on membranes. While the presence of Lysine residues is crucial for their activity, cardiotoxins demonstrate varying effects on different cells, indicating selective binding to certain cell types.

Application

Cardiotoxin, Naja pallida has been used:
  • to inject into the TA of the 4-month-old dystrophic mice to address whether ILA treatment could modulate muscle regeneration
  • to induce a skeletal muscle injury into the right TA muscle
  • to induce muscle regeneration

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Jang, J.Y., et al. 1997. Biochemistry 36, 4635.
Huang, J.L., and Trumble, W.R. 1991. Toxicon29, 31.
Raynor, R.L., et al. 1991. J. Biol. Chem.266, 2753.
Grognet, J.M., et al. 1986. Mol. Immunol.23, 132.

Legal Information

Not available for sale outside of the United States.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deficiency of skeletal muscle Agrin contributes to the pathogenesis of age-related sarcopenia in mice
Chen J, et al.
Cell Death & Disease, 15(3), 201-201 (2024)
Snake peptides
Harvey, Alan L
Handbook of Biologically Active Peptides (2013)
DOCK3 regulates normal skeletal muscle regeneration and glucose metabolism
Samani A, et al.
bioRxiv : the preprint server for biology (2023)
Evaluation of pro-regenerative and anti-inflammatory effects of isolecanoric acid in the muscle: Potential treatment of Duchenne Muscular Dystrophy
Matias-Valiente L, et al.
Biomedicine and Pharmacotherapy, 170, 116056-116056 (2024)
Valeria Yartseva et al.
Cell reports, 30(5), 1491-1503 (2020-02-06)
How satellite cells and their progenitors balance differentiation and self-renewal to achieve sustainable tissue regeneration is not well understood. A major roadblock to understanding satellite cell fate decisions has been the difficulty of studying this process in vivo. By visualizing expression

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service