Skip to Content
Merck
All Photos(1)

Key Documents

1.93636

Supelco

Calcium sulfate dihydrate

(precipitated), EMPLURA®

Synonym(s):

Calcium sulfate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CaO4S · 2H2O
CAS Number:
Molecular Weight:
172.17
MDL number:
UNSPSC Code:
12352302
form:
(precipitated)
Pricing and availability is not currently available.

product line

EMPLURA®

form

(precipitated)

storage temp.

room temp

SMILES string

[Ca+2].[S](=O)(=O)([O-])[O-].O.O

InChI

1S/Ca.H2O4S.2H2O/c;1-5(2,3)4;;/h;(H2,1,2,3,4);2*1H2/q+2;;;/p-2

InChI key

PASHVRUKOFIRIK-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service