Skip to Content
Merck
All Photos(2)

Key Documents

189513

Sigma-Aldrich

Autotaxin Inhibitor IV, HA155

The Autotaxin Inhibitor IV, HA155 controls the biological activity of Autotaxin. This small molecule/inhibitor is primarily used for Cell Structure applications.

Synonym(s):

Autotaxin Inhibitor IV, HA155, Atx Inhibitor IV, HA155, ( Z)-4-((4-((3-(4-Fluorobenzyl)-2,4-dioxo-1,3-thiazolan-5-yliden)-methyl)phenoxy)methyl)benzeneboronic acid, Atx Inhibitor IV, HA155, (Z)-4-((4-((3-(4-Fluorobenzyl)-2,4-dioxo-1,3-thiazolan-5-yliden)-methyl)phenoxy)methyl)benzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H19BFNO5S
CAS Number:
Molecular Weight:
463.29
MDL number:
UNSPSC Code:
41121800
NACRES:
NA.28

Quality Level

Assay

≥99% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

yellow

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(/C(S1)=C/C2=CC=C(OCC3=CC=C(B(O)O)C=C3)C=C2)N(CC4=CC=C(F)C=C4)C1=O

InChI key

BRWUZCBSWABPMR-XKZIYDEJSA-N

General description

A ~5-fold more potent (IC50 = 5.7 nM) para-isomer boronate analog of Autotaxin Inhibitor II, HA130 (Cat. No. 189511) that efficiently blocks thrombin-stimulated lysophosphatidic acid secretion in platelets and in mammalian cells.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Fulkerson, Z., et al. 2011. J. Biol. Chem.286, 34654.
Hausmann, J., et al. 2011. Nat. Struct. Mol. Biol.18, 198.
Albers, H.M.H.G., et al. 2011. J. Med. Chem.54, 4619.
Albers, H.M.H.G., et al. 2010. J. Med. Chem.53, 4958.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service