Skip to Content
Merck
All Photos(5)

Key Documents

17-10101

Sigma-Aldrich

ChIPAb+ Acetyl-Histone H4 (Lys16) - ChIP Validated Antibody and Primer Set

from rabbit

Synonym(s):

H4K16Ac, Histone H4 (acetyl K16), H4 histone family, member A, histone 1, H4a, histone cluster 1, H4a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.32

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

clone

polyclonal

species reactivity

vertebrates, human

manufacturer/tradename

ChIPAb+
Upstate®

technique(s)

ChIP: suitable
immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

All ChIPAb+ antibodies are individually validated for chromatin precipitation, every lot, every time. Each ChIPAb+ antibody set includes control primers (tested every lot by qPCR) to biologically validate your IP results in a locus-specific context. The qPCR protocol and primer sequences are provided, allowing researchers to validate ChIP protocols when using our antibody in their chromatin context. Each set also includes a negative control antibody to ensure specificity of the ChIP reaction.
The ChIPAb+ Acetyl-Histone H4 (Lys16) set includes the Acetyl-Histone H4 (Lys16) antibody, a negative control normal rabbit IgG, and qPCR primers which amplify a 178 bp region of human RPL10 promoter. The Acetyl-Histone H4 (Lys16) and negative controls are supplied in a scalable "per ChIP" reaction size and can be used to functionally validate the precipitation of Acetyl-Histone H4 (Lys16)-associated chromatin.

Specificity

Broad species cross-reactivity is expected based on sequence homology.
This antibody detects histone H4 acetylated at Lys16.

Immunogen

Epitope: Acetylated H4 Lys16
KLH-conjugated Histone H4 corresponding to the N-terminus region at and around acetylated Lys16.

Application

Chromatin Immunoprecipitation:
Representative lot data.
Sonicated chromatin prepared from HeLa cells (1 X 10E6 cell equivalents per IP) were subjected to chromatin immunoprecipitation using either 2 µg of Normal rabbit IgG or 2 µL of Anti-acetyl-Histone H4 (Lys16) and the Magna ChIP A Kit (Cat. # 17-610).
Successful immunoprecipitation of acetyl-Histone H4 (Lys16) associated DNA fragments was verified by qPCR using ChIP Primers, RPL10 Promoter as a positive locus, and β-Globin primers as a negative locus. (Please see figures). Data is presented as percent input of each IP sample relative to input chromatin for each amplicon and ChIP sample as indicated.
Please refer to the EZ-Magna ChIP A (Cat. # 17-408) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.

Western Blot Analysis:
Representative lot data.
Recombinant Histone H4 (Cat # 14-697) (lane1) and HeLa acid extract (lane 2) was resolved by electrophoresis, transferred to PVDF and probed with anti-Acetyl Histone H4 (Lys16) (0.1 μg/mL).
Proteins were visualized using a donkey anti-rabbit secondary antibody conjugated to HRP and a chemiluminescence detection system.
Arrow indicates Acetyl Histone H4 (Lys16) (~10 kDa) (Please see figures).

Multiplexing (Luminex):
Representative lot data.
This antibody specifically recognizes histone H4 acetylated on Lys16 by Luminex assay (Please see figures).

Peptide Inhibition Assay:
Representative lot data.
This antibody peptide blocked on HeLa cell extracts (Please see figures).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
This ChIPAb+ Acetyl-Histone H4 (Lys16) -ChIP Validated Antibody & Primer Set conveniently includes the antibody & the specific control PCR primers.

Packaging

25 assays per set. Recommended use: ~2 μL of antibody per chromatin immunoprecipitation (dependent upon biological context).

Quality

Chromatin Immunoprecipitation:
Sonicated chromatin prepared from HeLa cells (1 X 10E6 cell equivalents per IP) were subjected to chromatin immunoprecipitation using either 2 µg of Normal Rabbit IgG or 2 µL of Anti-acetyl-Histone H4 (Lys16) and the Magna ChIP® A Kit (Cat. # 17-610).
Successful immunoprecipitation of acetyl-Histone H4 (Lys16) associated DNA fragments was verified by qPCR using ChIP Primers, RPL10 Promoter (Please see figures).
Please refer to the EZ-Magna ChIP A (Cat. # 17-408) or EZ-ChIP (Cat. # 17-371) protocol for experimental details.

Target description

~10 kDa observed

Physical form

Affinity purified
Anti-Acetyl-Histone H4 (Lys16) (rabbit polyclonal). One vial containing 50 µL of affinity purified rabbit polyclonal serum in buffer containing 0.1 M Tris-Glycine (pH 7.4) 150 mM NaCl with 0.05% sodium azide. Store at -20°C.

Normal Rabbit IgG. One vial containing 125 µg of rabbit IgG in 125 µL of storage buffer containing 0.05% sodium azide. Store at -20°C.

ChIP Primers, RPL10 promoter. One vial containing 75 μL of 5 μM of each primer specific for the promoter region of human RPL10. Store at -20°C.
FOR: ACC CGT CTT CGA CAG GAC T
REV: GGA ACG GAA GAC GAG AAC AG
Format: Purified

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
Includes negative control normal rabbit IgG and primers specific for human RPL10 promoter.

Legal Information

MAGNA CHIP is a registered trademark of Merck KGaA, Darmstadt, Germany
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Umeshree Govender et al.
Journal of leukocyte biology, 101(5), 1181-1190 (2017-03-01)
Type I IFN can exert pro- and anti-inflammatory activities in the immune system. Here, we have investigated the mechanism by which IFN-α enhances early expression of the anti-inflammatory cytokine IL-10 in human CD45RA+CD4+ T cells. With the use of transcriptomic
Ye Zhang et al.
Journal of cell science, 131(12) (2018-05-16)
Dysregulation of the homeostatic balance of histone H3 di- and tri-methyl lysine 27 (H3K27me2/3) levels caused by the mis-sense mutation of histone H3 (H3K27M) is reported to be associated with various types of cancers. In this study, we found that
Keisuke Mori et al.
PloS one, 9(1), e87319-e87319 (2014-02-06)
We previously reported that sevoflurane anesthesia reversibly suppresses the expression of the clock gene, Period2 (Per2), in the mouse suprachiasmatic nucleus (SCN). However, the molecular mechanisms underlying this suppression remain unclear. In this study, we examined the possibility that sevoflurane
Hao Fu et al.
Molecular therapy oncolytics, 12, 235-245 (2019-03-09)
Clinical efficacy of redifferentiation therapy with histone deacetylase inhibitor (HDACi) for lethal radioiodine-refractory papillary thyroid cancer (RR-PTC) is urgently needed to be improved. Given that the impairment of histone acetylation is a mechanism in BRAF V600E-mitogen-activated protein kinase (MAPK)-induced aberrant
Zongjing Zhang et al.
Endocrine-related cancer, 21(2), 161-173 (2013-11-19)
The BRAF V600E mutation causes impaired expression of sodium iodide symporter (NIS) and radioiodine refractoriness of thyroid cancer, but the underlying mechanism remains undefined. In this study, we hypothesized that histone deacetylation at the NIS (SLC5A5) promoter was the mechanism.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service