17-10071
Magna GrIP HT96 Rack (for 96 well plates)
for use with separation of magnetic particles from solutions in a 96-well plate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104923
eCl@ss:
32011202
NACRES:
NB.51
Recommended Products
compatibility
for use with separation of magnetic particles from solutions in a 96-well plate
Quality Level
shipped in
ambient
General description
The Magna GrIP HT96 rack is designed for separation of magnetic particles from solutions in a 96-well plate, U-bottom microplates and 0.2 mL PCR plates with no additional accessories. This effective magnetic separation device is composed of white polyoxymethylene (POM) base in standard Society for Biomolecular Screening (SBS) 96-well microplate (12.8cm X 8.6cm) containing 24 (4 x 6) neodymium magnetic rods each having a diameter of approximately 7.5 mm and a Gauss rating of 6000 When a standard 96-well or PCR plate is fitted over the magnetic rack, each magnetic rod sits between four adjacent wells and rapidly pulls magnetic particles to the side or bottom of the wells
Application
Research Category
Epigenetics & Nuclear Function
Epigenetics & Nuclear Function
The Magna GrIP HT96 rack is designed for separation of magnetic particles from solutions in a 96-well plate.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service