Skip to Content
Merck
All Photos(1)

Documents

16-213

Sigma-Aldrich

Anti-Myc Tag Antibody, clone 4A6, HRP conjugate

clone 4A6, Upstate®, from mouse

Synonym(s):

Transcription factor p64, avian myelocytomatosis viral oncogene homolog, myc proto-oncogene protein, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.56

biological source

mouse

Quality Level

conjugate

peroxidase conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4A6, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... MYC(4609)

General description

Immunogen: peptide corresponding to amino acids 410-420 (MEQKLISEEDL) of human Myc

Specificity: Recognizes and is specific for recombinant proteins containing the Myc epitope tag (EQKLISEEDL) in a variety of sequence contexts. Also recognizes human Myc.

Species Cross-reactivity: Human, others not tested

Ig Type: IgG

Host: Mouse

Molecular Weight: Varies depending upon the protein which is tagged
Myc-Tag is a short synthetic peptide sequence derived from C-terminal region of human c-myc that is used as an epitope tag. The Myc tag can be cloned either upstream or downstream of the target gene, resulting in a fusion protein that allows efficient purification, detection, and localization of target proteins. The relatively small size of this tag vs. other commonly used tags such as GST (30 kDa) makes it less likely to interfere with target protein structure or function. The myc epitope tag is widely used to detect expression of recombinant proteins in bacterial, yeast, insect, and mammalian systems.

Specificity

Other species cross-reactivity not tested.
Recognizes and is specific for recombinant proteins containing the Myc epitope tag (EQKLISEEDL) in a variety of sequence contexts. Also recognizes human Myc.

Immunogen

Epitope: C-terminal
KLH-conjugated, synthetic peptide corresponding to amino acids 410-420 (MEQKLISEEDL) of human Myc. Clone 4A6.

Application

Anti-Myc Tag Antibody, clone 4A6, HRP conjugate detects level of Myc Tag & has been published & validated for use in WB.
Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags

Quality

Evaluated by Western Blot on Myc-PP2A A subunit transfected NIH/3T3 lysates.

Western Blot Analysis:
1:500 dilution of this antibody detected myc-PP2A on 10 µg of Myc-PP2A A subunit transfected NIH/3T3 lysates.

Target description

Varies depending upon the protein which is tagged.

Physical form

Protein G Purified
Purified mouse monoclonal IgG1 conjugated to horseradish peroxidase (HRP) in buffer containing: 50% storage buffer (PBS, 0.05% Kathon®, 0.2 mg/mL BSA) and 50% glycerol.

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.

Analysis Note

Control
Positive control for human Myc: Widespread expression including A431 cell lysate, ovarian cancer cell lysate, or breast carcinoma tissue. Myc fusion protein expressed in cells.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

Kathon is a registered trademark of Rohm and Haas Co.
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

390.2 °F - closed cup - Information refers to the main ingredient.

Flash Point(C)

199 °C - closed cup - Information refers to the main ingredient.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel transgenic mouse model of growth plate dysplasia reveals that decreased chondrocyte proliferation due to chronic ER stress is a key factor in reduced bone growth.
Gualeni, B; Rajpar, MH; Kellogg, A; Bell, PA; Arvan, P; Boot-Handford, RP; Briggs, MD
Disease models & mechanisms null
Catalytic assembly of the mitotic checkpoint inhibitor BubR1-Cdc20 by a Mad2-induced functional switch in Cdc20.
Han, JS; Holland, AJ; Fachinetti, D; Kulukian, A; Cetin, B; Cleveland, DW
Molecular Cell null
Junying Wang et al.
Nature communications, 15(1), 3481-3481 (2024-04-26)
Viral myocarditis, an inflammatory disease of the myocardium, is a significant cause of sudden death in children and young adults. The current coronavirus disease 19 pandemic emphasizes the need to understand the pathogenesis mechanisms and potential treatment strategies for viral
Raphael Trefzer et al.
PloS one, 16(5), e0252428-e0252428 (2021-05-28)
Diaphanous related formins are highly conserved proteins regulated by Rho-GTPases that act as actin nucleation and assembly factors. Here we report the functional characterization of a non-inherited heterozygous FMNL2 p.L136P mutation carried by a patient who presented with severe very
Hee Jun Cho et al.
Journal of microbiology and biotechnology, 30(10), 1488-1494 (2020-09-03)
Rho guanine nucleotide dissociation inhibitor 1 (RhoGDI1) plays important roles in numerous cellular processes, including cell motility, adhesion, and proliferation, by regulating the activity of Rho GTPases. Its expression is altered in various human cancers and is associated with malignant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service