Skip to Content
Merck
All Photos(1)

Documents

14-173-M

Sigma-Aldrich

MAP Kinase 2/Erk2 Protein, active, mouse, 10 µg

Active, N-terminal GST-tagged, recombinant MAP Kinase 2, expressed in E. coli cells, activated using MEK1 (Catalog #14-429). For use in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

biological source

mouse

Quality Level

recombinant

expressed in E. coli

mol wt

Mw 67.8 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

General description

N-terminal GST-tagged, recombinant MAP Kinase 2, expressed in E. coli cells; activated using MEK1 (Catalog #14-429).
Product Source: recombinant, mouse, full-length MAP Kinase 2

Biochem/physiol Actions

Protein Target: MAPK2
Target Sub-Family: CMGC

Packaging

Also available in 250 µg size --call for pricing and availability and reference catalog number 14-173M when ordering the 250 µg size.

Quality

Routinely evaluated by phosphorylation myelin basic protein

Physical form

50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.1mM EGTA, 0.03% Brij-35, 50% glycerol, 1mM benzamidine, 0.2mM PMSF, 0.1% 2-mercaptoethanol.
Glutathione agarose; Ni2+/NTA agarose

Storage and Stability

Store in undiluted aliquots at -20°C for up to one year from the date of receipt. Avoid repeatede freeze-thaw cycles.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tyrosyl phosphorylation and activation of the myelin basic protein kinase p44mpk during sea star oocyte maturation.
Sanghera, J S, et al.
Biochimica et Biophysica Acta, 1095, 153-160 (1991)
alpha7 Nicotinic Acetylcholine Receptors Mediate {beta}-Amyloid Peptide-induced Tau Protein Phosphorylation
Wang, H. Y., et al
The Journal of Biological Chemistry, 278, 31547-31553 (2003)
B Zhang et al.
The Journal of biological chemistry, 271(50), 31771-31774 (1996-12-13)
Peroxisome proliferator-activated receptor (PPAR) gamma plays an important role in adipocyte differentiation and the regulation of adipocyte gene expression. Insulin also serves to promote adipogenesis. We report that insulin and a PPARgamma ligand (thiazolidinedione (TZD)) stimulate in a synergistic manner
Purification and characterization of a maturation-activated myelin basic protein kinase from sea star oocytes.
Sanghera, J S, et al.
The Journal of Biological Chemistry, 265, 52-57 (1990)
X Yang et al.
The Journal of biological chemistry, 273(45), 29879-29887 (1998-10-29)
The human immunodeficiency virus type 1 (HIV-1) Vif protein plays a critical role in virus replication and infectivity. Here we show that Vif is phosphorylated and regulated by p44/42 mitogen-activated protein kinase (MAPK). Vif phosphorylation by MAPK was demonstrated in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service