Skip to Content
Merck
All Photos(1)

Key Documents

12-506

Sigma-Aldrich

Goat Anti-Mouse IgG (H+L) Antibody, FITC conjugate

Upstate®, from goat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.42

biological source

goat

Quality Level

conjugate

FITC conjugate

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Upstate®

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

shipped in

dry ice

target post-translational modification

unmodified

Specificity

Recognizes mouse IgG, both heavy and light chains. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-fluorescein, anti-goat serum, mouse IgG and mouse serum.

Immunogen

Mouse IgG whole molecule.

Application

Absorption Wavelength: 495nm
Emission Wavelength: 528nm
Fluorochrome/Protein Molar Ratio: 3.2

Suitable for immunomicroscopy and flow cytometry or FACS analysis, as well as other antibody based fluorescent assays requiring lot-to-lot consistency. Optimal antibody dilution should be determined by the researcher.
Detect Mouse IgG (H+L) using this Goat anti-Mouse IgG (H+L) Antibody, FITC conjugate validated for use in FC, IF, IC, IH.
Research Category
Secondary & Control Antibodies
Research Sub Category
Whole Immunoglobulin Secondary Antibodies

Quality

Routinely evaluated by immunomicroscopy, flow cytometry or FACS analysis, as well as other antibody based fluorescent assays requiring lot-to-lot consistency.

Physical form

ImmunoAffinity Purified
Lyophilized powder; lyophilized from 500 μl of 20 mM potassium phosphatase, pH 7.2, 150 mM NaCl, 0.10% sodium azide and 10 mg/ml BSA.

Storage and Stability

Stable for 1 year at -20°C from date of shipment. Once opened, dilute with 250μl of distilled water, aliquot, and freeze at -20°C or below for extended storage. Avoid multiple freeze-thaw cycles. Dilute only immediately prior to use.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Differential regulation of lipogenesis and leptin production by independent signaling pathways and rosiglitazone during human adipocyte differentiation.
Nayan G Patel,Julie C Holder,Stephen A Smith,Sudesh Kumar,Margaret C Eggo
Diabetes null
Na Wang et al.
Cellular oncology (Dordrecht), 44(5), 1119-1131 (2021-09-13)
Intestinal metaplasia (IM) is a precancerous lesion that increases the risk of subsequent gastric cancer (GC) development. Previously, miR-1 has been shown to play an essential role in the initiation of bile acid (BA)-induced IM. The objective of the present
Effect of tacrolimus on chemokine production by corneal myofibroblasts via toll-like receptors, compared with cyclosporine and dexamethasone.
Nobuyuki Ebihara,Kaori Ohtomo,Tomoko Tokura,Hiroko Ushio,Akira Murakami
Cornea null
Malick Bio Idrissou et al.
Pharmaceutics, 13(7) (2021-07-03)
Auger electron emitters (AEEs) are attractive tools in targeted radionuclide therapy to specifically irradiate tumour cells while sparing healthy tissues. However, because of their short range, AEEs need to be brought close to sensitive targets, particularly nuclear DNA, and to
Rapamycin inhibits human adipocyte differentiation in primary culture.
A Bell,L Grunder,A Sorisky
Obesity Research null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service