Skip to Content
Merck
All Photos(1)

Key Documents

1.19814

Supelco

Fluoride standard solution

Fluoride standard solution
1 of 1 reviewers received a sample product or took part in a promotion

traceable to SRM from NIST NaF in H₂O 1000 mg/l F Certipur®

Synonym(s):

Fluoride standard for aquecous matrices ready for use

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

Quality Level

technique(s)

ion chromatography: suitable

pH

6 (20 °C in H2O)

density

1.000 g/cm3 at 20 °C

application(s)

environmental
food and beverages

format

single component solution

storage temp.

15-25°C

Application


  • Fluoride Calibration Standard: Utilized extensively in biochemical fluoride analysis, this solution ensures precise calibration of analytical instruments, crucial for accurate fluoride ion quantification in dental and pharmaceutical research (Kallitsis et al., 2024).

  • Fluoride Assay Reagent: This high-purity fluoride reference solution is a key reagent in various biochemical assays, helping to study fluoride′s role in inhibiting enzymes that are linked to dental and bone diseases (Liu et al., 2023).

  • Biochemical Fluoride Analysis: In pharmaceutical and biochemical labs, this fluoride standard is used for assays that determine fluoride′s therapeutic potential and toxicity, providing essential data for drug development and safety evaluations (Bui et al., 2023).

  • Antifungal Drug Discovery: This standard solution is also used in antifungal drug research, particularly in studying fluoride′s role as a potential treatment for fungal infections by inhibiting fungal enzyme systems (Li et al., 2023).

Analysis Note

Concentration     β (F⁻): 990 - 1010 mg/l
Determination method: Titration with Lanthannitrate solution
(traceable to NIST SRM 682)
Accuracy of the method: ± 2 mg/l

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. 1 Ratings-only review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service