Skip to Content
Merck
All Photos(1)

Key Documents

1.19159

Supelco

Standard samples for HPLC system qualification

suitable for HPLC, parabens in methanol/water, for use with LICHROTEST

Synonym(s):

TM=["LICHROTEST"] Standard samples for HPLC system qualification Set 3: Separation

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
NACRES:
NA.24

grade

analytical standard

Quality Level

composition

parabens in methanol/water

technique(s)

HPLC: suitable

compatibility

for use with LICHROTEST

format

mixture

storage temp.

15-25°C

General description

Set 3: Separation; Parabens in methanol / water (50:50) LiChroTest

Analysis Note

1 mg/l Thiourea: in accordance
10 mg/l Methyl paraben: in accordance
10 mg/l Ethyl paraben: in accordance
10 mg/l Propyl paraben: in accordance

Legal Information

LICHROTEST is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service